Skip to Content
Merck
All Photos(1)

Documents

PHR1385

Supelco

Ketoconazole

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ketoconazole, (±)-cis-1-Acetyl-4-(4-[(2-[2,4-dichlorophenyl]-2-[1H-imidazol-1-ylmethyl]-1,3-dioxolan-4-yl)-methoxy]phenyl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28Cl2N4O4
CAS Number:
Molecular Weight:
531.43
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. K0600000
traceable to USP 1356508

API family

ketoconazole

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC(=O)N1CCN(CC1)c2ccc(OC[C@H]3CO[C@@](Cn4ccnc4)(O3)c5ccc(Cl)cc5Cl)cc2

InChI

1S/C26H28Cl2N4O4/c1-19(33)31-10-12-32(13-11-31)21-3-5-22(6-4-21)34-15-23-16-35-26(36-23,17-30-9-8-29-18-30)24-7-2-20(27)14-25(24)28/h2-9,14,18,23H,10-13,15-17H2,1H3/t23-,26-/m0/s1

InChI key

XMAYWYJOQHXEEK-OZXSUGGESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Ketoconazole is an imidazole antifungal agent administered through topical or oral means. It is used for the treatment of chronic mucocutaneous candidiasis, fungal infections of the gastro-intestinal tract, dermatophyte infections, systemic infections, and fungal infections in immuno-compromised patients.

Application

Ketoconazole may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
CYP3A4 inhibitor

Biochem/physiol Actions

Antifungal agent

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3465 in the slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of capillary zone electrophoresis and high performance liquid chromatography methods for quantitative determination of ketoconazole in drug formulations
Velikinac I, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 59(5), 419-424 (2004)
R A Bok et al.
Drug safety, 20(5), 451-458 (1999-05-29)
The definition of hormone refractory prostate cancer is changing. It has become clear that patients with advanced prostate cancer whose disease has progressed following treatment with luteinising hormone releasing hormone agonists and antiandrogens can respond to additional hormonal manoeuvres. Ketoconazole
Jan Faergemann et al.
Expert opinion on pharmacotherapy, 8(9), 1365-1371 (2007-06-15)
Seborrhoeic dermatitis (SD) is a chronic, inflammatory skin disorder, affecting areas of the head and body where sebaceous glands are most prominent and active. The disorder commonly affects hair-bearing areas of the head, including the scalp. Involvement on the face
Chromatographic determination of clotrimazole, ketoconazole and fluconazole in pharmaceutical formulations
Abdel-Moety EM, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 57(11), 931-938 (2002)
J J Berwaerts et al.
Journal of the American Geriatrics Society, 46(7), 880-884 (1998-07-22)
Cushing's syndrome is a rare disorder. The corticotropin (ACTH)-dependent form of this syndrome generally results either from excessive ACTH secretion by a pituitary adenoma or ectopic secretion by a malignant tumor. Theoretically, the latter type can be assumed to occur

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service