Skip to Content
Merck
All Photos(1)

Documents

1061901

USP

Benzyl alcohol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(Hydroxymethyl)benzene, Phenylmethanol, Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

API family

benzalkonium chloride

autoignition temp.

817 °F

shelf life

~3 yr

manufacturer/tradename

USP

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzyl alcohol USP reference standard meant for use in specified quality tests and assays.

Also used to prepare standard preparation, system suitability, standard and system suitability stock solution for assay and impurity analysis according to the given below monographs of United States Pharmacopeia (USP):
  • Midazolam Injection
  • Famotidine Injection
  • Enalaprilat Injection
  • Cisatracurium Besylate Injection
  • Physostigmine Salicylate Injection
Additionally, it is used to prepare an internal standard solution by using gas chromatography in conjunction with a flame ionization detector (GC-FID) according to the United States Pharmacopeia (USP)chapter.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physostigmine Salicylate Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 30(6), 3535-3535 (2018)
Cisatracurium Besylate Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(5), 1028-1028 (2020)
Midazolam Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 34(3), 2953-2953 (2018)
Enalaprilat Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 34(3), 1619-1619 (2022)
Famotidine Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(3), 1816-1816 (2018)

Related Content

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service