Skip to Content
Merck
All Photos(1)

Key Documents

1.02695

Supelco

Benzyl alcohol

for headspace gas chromatography SupraSolv®

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
MDL number:
UNSPSC Code:
41116105
EC Index Number:
202-859-9
NACRES:
NA.03
Assay:
≥99.5% (GC)
technique(s):
GC-HS: suitable
bp:
203-205 °C (lit.)
vapor pressure:
0.07 hPa ( 20 °C)
13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)
Pricing and availability is not currently available.

Agency

according to Ph. Eur.
according to USP

Quality Level

vapor density

3.7 (vs air)

vapor pressure

0.07 hPa ( 20 °C)
13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

product line

SupraSolv®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

435 °C
817 °F

technique(s)

GC-HS: suitable

impurities

≤0.001 meq/g Acidity
≤0.002 meq/g Alkalinity
≤0.1% Water

color

APHA: ≤10

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

transition temp

flash point 101 °C

density

1.045 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv® high-purity solvents for headspace-gas chromatography (HS-GC) are developed specially for the analysis of residual solvents (RS) in active pharmaceutical ingredients (APIs), excipients and drug products according to ICH (International Council for Harmonizaton of Technical Requirements for Registration of Pharmaceuticals for Human Use) Guideline Q3C “Impurities: Guideline for Residual Solvents”.

Application

Solvent for HS-GC analysis of residual solvents of ICH classes 1, 2 and 3 according to Ph. Eur. And USP.

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Colour: ≤ 10 Hazen
Acidity: ≤ 0.001 meq/g
Alkalinity: ≤ 0.002 meq/g
Water: ≤ 0.1 %
GC/HS (Every residual solvent of class 1acc. ICH): ≤ 1 µg/g
GC/HS (Every residual solvent of class 2acc. ICH): ≤ 10 µg/g
GC/HS (Every residual solvent of class 3acc. ICH): ≤ 50 µg/g
Suitable as solvent for the analysis of residual solvents of ICH classes 1, 2 and 3 according to Ph Eur and USP.
Each unknown measured against butyl methyl ketone up to anisole is ≤ 10 µg/g.
Formic acid, acetic acid, dimethylacetamide and dimethyl sulfoxide are not detected.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service