Skip to Content
Merck
All Photos(4)

Key Documents

402834

Sigma-Aldrich

Benzyl alcohol

ACS reagent, ≥99.0%

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
EC Number:
MDL number:
UNSPSC Code:
12352104
eCl@ss:
39023110
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
Assay:
≥99.0%
bp:
203-205 °C (lit.)
vapor pressure:
13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

817 °F

expl. lim.

0.34-6.3 %

impurities

≤0.01% Benzaldehyde (C6H5CHO)
≤0.02% Acetophenone (C6H5COCH3)

ign. residue

≤0.005%

color

APHA: ≤20

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl alcohol (BnOH) is a monoaromatic primary alcohol widely used as a solvent and an intermediate in cosmetic formulations, pharmaceutical and flavor/fragrance industries. It shows bacteriostatic property. Commercial method for its synthesis is by the alkaline hydrolysis of benzyl chloride.[1] Conformational studies by ab initio calculations shows gauche cis conformation around the alcoholic group.[2]

Application

Benzyl alcohol may be used for the benzylation of toluene.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The most stable conformer of benzyl alcohol.
Miller BJ, et al.
Chemical Physics Letters, 466(1), 21-26 (2008)
Engineering Escherichia coli for renewable benzyl alcohol production.
Pugh S, et al.
Metabolic Engineering Communications, 2, 39-45 (2015)
Application of statistical design to benzylation of toluene with benzyl alcohol.
Alam A, et al.
Chemical Engineering & Technology, 31(3), 417-420 (2008)
Poincyane Assis-Nascimento et al.
Journal of neuroscience methods, 263, 57-67 (2016-02-09)
Traumatic brain injury (TBI) continues to be a major source of death and disability worldwide, and one of the earliest and most profound deficits comes from vascular damage and breakdown of the blood-brain barrier (BBB). Cerebral vascular endothelial cells (cvECs)
Po-Yu Liao et al.
Frontiers in neuroanatomy, 14, 559402-559402 (2020-11-17)
Oxytocin, a neuropeptide and peptide hormone, is produced by neurons in the hypothalamus and released by the posterior pituitary to control breastfeeding and labor. Recent studies have revealed that oxytocin in the central nervous system is also involved in modulating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service