Skip to Content
Merck
All Photos(1)

Key Documents

1048506

USP

Beclomethasone dipropionate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37ClO7
CAS Number:
Molecular Weight:
521.04
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

beclometasone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C

InChI

1S/C28H37ClO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1

InChI key

KUVIULQEHSCUHY-XYWKZLDCSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Beclomethasone dipropionate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Beclomethasone Dipropionate Compounded Oral Solution
  • Betamethasone Dipropionate
  • Betamethasone Dipropionate Cream
  • Betamethasone Dipropionate Lotion
  • Betamethasone Dipropionate Ointment
  • Betamethasone Valerate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Adrenal gland,Immune system,Bone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficacy of inhaled beclomethasone dipropionate and budesonide in the treatment of asthma.
D R Webb
Respiratory medicine, 92 Suppl B, 7-14 (1999-04-08)
Comparative review of the effects of inhaled beclomethasone dipropionate and budesonide on bone.
M D'Souza
Respiratory medicine, 92 Suppl B, 24-36 (1999-04-08)
Effect of inhaled beclomethasone dipropionate and budesonide on adrenal function, skin changes and cataract formation.
R G Dluhy
Respiratory medicine, 92 Suppl B, 15-23 (1999-04-08)
J C Guérin et al.
Presse medicale (Paris, France : 1983), 21(32), 1526-1531 (1992-10-03)
Beclomethasone dipropionate has now been used for more than 10 years during which our knowledge of how to use inhaled corticosteroids has gradually improved: high dose initial treatment followed by progressive reduction down to the minimum effective dosage; administration in
T B Edwards
Clinical therapeutics, 17(6), 1032-1041 (1995-11-01)
There is an increasing trend toward topical intranasal corticosteroids as the preferred treatment for seasonal or perennial allergic rhinitis. This trend is based on the fact that the chronic, mucosal inflammation that accompanies allergic rhinitis responds to anti-inflammatory drugs such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service