Skip to Content
Merck
All Photos(1)

Documents

1044403

USP

Atenolol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide, 4-[2′-Hydroxy-3′-(isopropylamino)propoxy]phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O3
CAS Number:
Molecular Weight:
266.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

atenolol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)NCC(O)COc1ccc(CC(N)=O)cc1

InChI

1S/C14H22N2O3/c1-10(2)16-8-12(17)9-19-13-5-3-11(4-6-13)7-14(15)18/h3-6,10,12,16-17H,7-9H2,1-2H3,(H2,15,18)

InChI key

METKIMKYRPQLGS-UHFFFAOYSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Atenolol USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Atenolol Tablets
  • Atenolol Injection
  • Atenolol Compounded Oral Suspension
  • Atenolol and Chlorthalidone Tablets
  • Atenolol Compounded Oral Suspension, Veterinary
  • Levobunolol Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Atenolol: pharmacokinetic/dynamic aspects of comparative developmental toxicity.
Sonia A Tabacova et al.
Reproductive toxicology (Elmsford, N.Y.), 16(1), 1-7 (2002-04-06)
H Sakurai et al.
Japanese circulation journal, 64(11), 893-896 (2000-12-08)
Cardiogenic shock developed in a 72-year-old Japanese woman during combination therapy with verapamil and atenolol for recurrent supraventricular arrhythmia. She had coronary atherosclerosis, liver cirrhosis and bradycardia-tachycardia syndrome. Despite of the high-dose catecholamines and counterpulsation, she progressively deteriorated. Bolus administration
Ronald V Lacro et al.
The New England journal of medicine, 371(22), 2061-2071 (2014-11-19)
Aortic-root dissection is the leading cause of death in Marfan's syndrome. Studies suggest that with regard to slowing aortic-root enlargement, losartan may be more effective than beta-blockers, the current standard therapy in most centers. We conducted a randomized trial comparing
M McGuiness et al.
Journal of the American Academy of Dermatology, 37(2 Pt 2), 298-299 (1997-08-01)
Atenolol is a beta-blocker commonly used for treating hypertension. It can induce various kinds of adverse side effects, including psoriasiform skin eruptions, skin necrosis, vasculitis, and (rarely) drug-induced connective tissue disease. We encountered a patient receiving atenolol for his hypertension
Laura M Kuyper et al.
The Canadian journal of cardiology, 30(5 Suppl), S47-S53 (2014-04-23)
Previous reviews have shown that β-blocker use for the treatment of hypertension without compelling indications was associated with increased risk of stroke in the elderly. It remains unclear whether this increased risk was driven by the type of β-blocker. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service