Skip to Content
Merck
All Photos(1)

Key Documents

Y0000352

Beclometasone dipropionate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Beclomethasone dipropionate, 9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37ClO7
CAS Number:
Molecular Weight:
521.04
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

beclometasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C

InChI

1S/C28H37ClO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1

InChI key

KUVIULQEHSCUHY-XYWKZLDCSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Beclometasone dipropionate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Adrenal gland,Immune system,Bone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Chervinsky
Clinical therapeutics, 18(5), 790-796 (1996-09-01)
A new double-strength (84 micrograms/spray) formulation of beclomethasone dipropionate (BDP-ds) as an aqueous suspension has been introduced to control symptoms of allergic rhinitis with once-daily intranasal dosing. This paper reviews the results of three clinical reports which show that BDP-ds
R N Brogden et al.
Drugs, 28(2), 99-126 (1984-08-01)
Inhaled beclomethasone dipropionate is now well established in the management of asthma. Studies conducted over the last decade, and since the drug was previously reviewed in the Journal, have confirmed that inhaled beclomethasone dipropionate 400 to 800 micrograms daily can
B Davies
The British journal of clinical practice, 47(2), 87-93 (1993-03-01)
Several reports have claimed that beclomethasone dipropionate (BDP) and budesonide (BUD) differ with regard to therapeutic efficacy and associated adverse effects, based either upon clinical data, or upon theoretical considerations. Although long-term comparisons are lacking, the consensus of numerous studies
S E Libretto
Archives of toxicology, 69(8), 509-525 (1995-01-01)
This paper reviews the published toxicity of beclomethasone dipropionate (BDP). BDP is a synthetic glucocorticosteroid which has a powerful local anti-inflammatory effect but little systemic action. It has been developed for both dermatological and inhaled applications. LD50 values and other
A R Boobis
Respiratory medicine, 92 Suppl B, 2-6 (1999-04-08)
The physicochemical and pharmacokinetic characteristics of BDP and budesonide are somewhat different, but the overall result is that both are well suited for use as inhaled corticosteroids. Both BDP and budesonide are metabolized primarily by the liver, with one of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service