Skip to Content
Merck
All Photos(1)

Documents

1031503

USP

Amoxicillin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Amoxicillin trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O5S · 3H2O
CAS Number:
Molecular Weight:
419.45
Beilstein:
7507120
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

amoxicillin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccc(O)cc3)C(=O)N2[C@H]1C(O)=O

InChI

1S/C16H19N3O5S.3H2O/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7;;;/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24);3*1H2/t9-,10-,11+,14-;;;/m1.../s1

InChI key

MQXQVCLAUDMCEF-CWLIKTDRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amoxicillin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Amoxicillin Tablets
  • Amoxicillin Capsules
  • Amoxicillin Boluses
  • Amoxicillin Oral Suspension
  • Amoxicillin Intramammary Infusion
  • Amoxicillin for Injectable Suspension

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amoxicillin
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(3), 297-297 (2020)
Amoxicillin Capsules
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(6), 300-300 (2013)
Amoxicillin Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(6), 304-304 (2021)
Amoxicillin Boluses
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 299-299 (2013)
Tony Antoniou et al.
CMAJ : Canadian Medical Association journal = journal de l'Association medicale canadienne, 187(4), E138-E143 (2015-02-04)
Trimethoprim-sulfamethoxazole increases the risk of hyperkalemia when used with spironolactone. We examined whether this drug combination is associated with an increased risk of sudden death, a consequence of severe hyperkalemia. We conducted a population-based nested case-control study involving Ontario residents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service