Skip to Content
Merck
All Photos(1)

Documents

A1300000

Ascorbic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Ascorbic acid, L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP Reference standard

API family

ascorbic acid

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

190-194 °C (dec.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ascorbic EP reference standard, intended for use only as specifically prescribed in the European Pharmacopoeia. Also used for the determination of the analyte in pharmaceutical formulations by chromatography and spectrophotometric techniques.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reversed-phase ion-pair HPLC determination of some water-soluble vitamins in pharmaceuticals
Ivanovic D, et al.
Journal of Pharmaceutical and Biomedical Analysis, 18(6), 999-1004 (1999)
Ascorbic acid
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 18(6), 6868-6869 (2019)
Spectrophotometric determination of L-ascorbic acid in pharmaceuticals
Szpikowska-Sroka B and Poledniok J
Journal of Analytical Chemistry, 66(10), 941-941 (2011)
Vitamin C: One compound, several uses. Advances for delivery, efficiency and stability
Carita AC, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 31(2), 102117-102117 (2019)
Vitamin c
Linster CL and Van SE
FEBS Journal, 274(1), 1-22 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service