RAD405
radiello™ BTEX Calibration Kit (CS2 Desorption)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NB.24
Recommended Products
application(s)
environmental
format
mixture
General description
The BTEX Calibration Kit (CS2 Desorption) was designed for the analysis of BTEX in urban environments. The mass of each compound loaded onto each cartridge span the extreme ranges represented in most urban environments. The kit may be used for both routine calibration and quality control. The kit consists of twelve RAD130 Cartridge Adsorbents. Three of which are blanks and nine, divided into three concentration groups, are preloaded with BTEX to simulate 7 day exposures (100,080 minutes). Concentrations are described in the listed table. The values shown are indicative. Actual concentrations are certified for each lot. The cartridges are stable for at least four months when stored at 4°C.
Legal Information
radiello is a trademark of Institi Clinici Scientifici Maugeri
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Isabel M Morales Terrés et al.
Journal of environmental management, 91(12), 2754-2762 (2010-09-03)
This paper describes a novel methodology for evaluating the extent to which petrol stations affect their surroundings. The method is based on the fact that the ratio of the concentrations of aliphatic and aromatic hydrocarbon pollutants in the air of
C Minoia et al.
The Science of the total environment, 192(2), 163-182 (1996-12-02)
A study using individual dosimetry to evaluate the daily inhaled dose of sixteen aromatic and aliphatic hydrocarbons in three groups of primary school children, living in three Italian towns with 50,000 inhabitants or less, (Treviglio-Lombardy; Poggibonsi-Tuscany; Valenza-Piedmont) is presented. The
Study of the dispersion of VOCs emitted by a municipal solid waste landfill
Chiriac, R., et al.
Atmospheric Environment, 43 (11), 1926-1931 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service