The lot specific Certificate of Analysis gives detailed information on the analytical method parameters. Please access a Certificate of Analysis in the DOCUMENTATION section under 'Certificate of Analysis':
https://www.sigmaaldrich.com/product/supelco/crm48777#product-documentation
CRM48777
EPA VOC Mix 2
certified reference material, 2000 μg/mL each component in methanol, ampule of 1 mL
Synonym(s):
EPA VOC Mix 2
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
CofA
current certificate can be downloaded
packaging
ampule of 1 mL
concentration
2000 μg/mL each component in methanol
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
multi-component solution
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Analyte
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1A - Flam. Liq. 2 - Muta. 1B - STOT SE 1
Target Organs
Eyes,Central nervous system
WGK
WGK 3
Flash Point(F)
closed cup
Flash Point(C)
closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
does the calibration mix come with suggested method parameters for testing?
1 answer-
Helpful?
-
-
CRM の標準物質が必要で、EPA VOC Mix 2 の試験成績書の見本をダウンロードしてみました。そこには不確かさの記載はあったのですが、トレーサビリティーチャートは付いていませんでした。製品を購入した場合は、トレーサビリティーチャートも付属するのでしょうか?
1 answer-
More recent lots offer a reformatted Certificate of Analysis. Note that this product offers metrological traceability which is traceable to the SI and higher order standards from NIST through an unbroken chain of comparisons. The balance used to weigh raw materials is accurate to +/-0.0001 g and calibrated regularly using mass standards traceable to NIST. Additionally, individual analytes are traceable to NIST SRMs where available. Please see the link below to review the Certificate of Analysis for a more recent lot:
https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/rtc/CRM48777_LRAC9820.pdfTo discuss further, we kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquires" under the Products Section with all the required information so that a member of our team can reach out to you to assist further.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service