Skip to Content
Merck
All Photos(1)

Key Documents

180173

Sigma-Aldrich

Carbon disulfide

ACS reagent, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
Beilstein:
1098293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
46 °C (lit.)
vapor pressure:
5.83 psi
Pricing and availability is not currently available.

grade

ACS reagent

vapor density

2.67 (vs air)

vapor pressure

5.83 psi

Assay

≥99.9%

form

liquid

autoignition temp.

212 °F

expl. lim.

50 %

dilution

(for analytical testing)

impurities

H2S, passes test (lim. ~1.5 ppm)
SO2, passes test (lim ~2.5 ppm)
≤0.05% water

evapn. residue

≤0.002%

color

APHA: ≤10

refractive index

n20/D 1.627 (lit.)

bp

46 °C (lit.)

mp

−112-−111 °C (lit.)

density

1.266 g/mL at 25 °C (lit.)

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carbon disulfide (CS2) is a colorless liquid. It is present in atmosphere and natural waters in the form of sulfur gas. It is responsible for the production of sulfur dioxide (SO2) in the atmosphere (stratosphere and/or troposphere).[1] Its lifetime in the atmosphere has been proposed to be around 12 days.[2] It is a non-polar solvent.[3]
Carbon disulfide is a highly volatile, flammable liquid with low ignition temperature. It can be synthesized by reacting hydrocarbon gas with sulfur. It is an important raw material for preparing viscose rayon and cellophane film.[4]

Application

Carbon disulfide has been used for the desorption of air samples during analysis of trihalomethanes (THMs) in those samples.[5]
It may be used in the preparation of dibenzyl trithiocarbonate, a RAFT (Reversible Addition-Fragmentation Transfer) chain-transfer agent (CTA).[6] It may be employed as a solvent in the preparation of benzyl chlorodithioformate.[7]
Carbon disulfide may be used in the xanthogenation of cellulose[8] and in the synthesis of poly(ethylene trithiocarbonate)[9] and dithiocarbamates.[10] C2S may undergo hydrolysis catalyzed by nanosized titania and zirconia to form CO2 and H2S.[11] It can also generate 1,3-dithiolium carbenes via cycloaddition with acetylenes.[12]

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copolymerization of ethylene sulfide and carbon disulfide.
Soga K, et al.
Journal of Polymer Science, 14(3), 677-684 (1976)
1, 3-Dithiolium carbenes from acetylenes and carbon disulfide.
Hartzler HD.
Journal of the American Chemical Society, 95(13), 4379-4387 (1973)
Nanosized titania and zirconia as catalysts for hydrolysis of carbon disulfide.
Yue Y, et al.
Applied Catalysis. B, Environmental, 46(3), 561-572 (2003)
Carbon Disulfide.
Smith DE and Timmerman RW.
Kirk-Othmer Encyclopedia of Chemical Technology (2003)
Cyril Catto et al.
International journal of environmental research and public health, 9(8), 2562-2586 (2012-10-16)
In order to improve disinfection by-product (DBP) exposure assessment, this study was designed to document both water and air levels of these chemical contaminants in two indoor swimming pools and to analyze their within-day and day-to-day variations in both of

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service