Skip to Content
Merck
All Photos(1)

Key Documents

58355-U

Supelco

SUPELCOSIL LC-18-DB (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Synonym(s):

High Purity C18 HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

SUPELCOSIL LC-18-DB HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L1

product line

SUPELCOSIL

feature

endcapped

manufacturer/tradename

SUPELCOSIL

packaging

1 ea of

extent of labeling

11.0% Carbon loading

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.1 μmol/m2

matrix

silica gel, spherical particle platform
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

operating pH

2-7.5

application(s)

food and beverages

compatibility

for use with PTC-AA

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL LC-DB phases are specially deactivated for basic compounds. These columns provide shorter retention, better peak shape, and higher efficiency for organic bases than can be obtained on other Type A silica reversed-phase columns.

Application


  • OPTIMIZATION AND VALIDATION OF HPLC METHOD FOR TETRAMETHRIN DETERMINATION IN HUMAN SHAMPOO FORMULATION.: This study discusses the optimization and validation of a high-performance liquid chromatography (HPLC) method using SUPELCOSIL LC-18-DB HPLC Column for the analysis of tetramethrin in shampoo formulations (Zeric Stosic et al., 2016).

  • Mass spectrometric characterization and HPLC determination of the main urinary metabolites of nimesulide in man.: Details a robust approach for identifying and quantifying nimesulide metabolites via HPLC, underscoring the column′s applicability in complex biological matrices (Carini et al., 1998).


Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D C Holland et al.
Journal of AOAC International, 78(4), 1067-1071 (1995-07-01)
A liquid chromatographic (LC) method is described for the simultaneous determination of the triazine herbicides, simazine (SIM), atrazine (ATZ), and propazine (PRO) in the 12.5-100 ppb range in catfish. The herbicides are extracted from catfish homogenates with ethyl acetate, followed
P A Hynning et al.
Clinical chemistry, 34(12), 2502-2503 (1988-12-01)
A high-performance liquid chromatographic (HPLC) method for determining verapamil and norverapamil in plasma is presented and compared with gas chromatography/mass spectrometry (GC-MS). The plasma samples were extracted at alkaline pH with hexane containing 2-butanol (20 mL/L) and then back-extracted into
M Carini et al.
Journal of pharmaceutical and biomedical analysis, 18(1-2), 201-211 (1998-12-24)
A study was undertaken for the characterization and quantitative determination of the main urinary metabolites of the non-steroidal anti-inflammatory drug (NSAID) nimesulide (4-nitro-2-phenoxy-methanesulfonanilide) in man following single oral administration (200 mg). Urines were collected from six healthy volunteers at 12
O Majid et al.
Therapeutic drug monitoring, 23(2), 163-168 (2001-04-11)
Recipients of organ transplants remain particularly dependent on prednisolone as part of their maintenance immunosuppression. Despite this, the pharmacokinetics of prednisolone have never been fully characterized in these patients, and consequently dosing remains empirical. Accurate monitoring of prednisolone, its primary
Xinghua Sun et al.
Anticancer research, 25(1A), 59-62 (2005-04-09)
A rapid, sensitive and specific high-performance liquid chromatographic (HPLC) method for the separation and quantification of L-methionine in plasma has been developed. After derivatization of plasma amino acids with o-phthalaldehyde (OPA), a 50 microl sample was loaded on a reversed-phase

Protocols

Amino Acid Analysis is the suitable tool for precise determination of protein quantities but also provides detailed information regarding the relative amino.

Chromatograms

application for SPE, application for HPLC

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service