Skip to Content
Merck
All Photos(1)

Documents

49156

Supelco

EPA TCL PAH Mix

certified reference material, in acetonitrile: methanol (9:1) (varied)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 8310,IP-7,550.1,551.1,TO-13,TCL
NIOSH 5506,5515

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in acetonitrile: methanol (9:1) (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthene 1000 μg/mL
    Acenaphthylene 500 μg/mL
    Anthracene 20 μg/mL
    Benz[a]anthracene 50 μg/mL
    Benzo[b]fluoranthene 20 μg/mL
    Benzo[k]fluoranthene 20 μg/mL
    Benzo[ghi]perylene 80 μg/mL
    Benzo[a]pyrene 50 μg/mL
    Chrysene 50 μg/mL
    Dibenz[a,h]anthracene 200 μg/mL
    Fluoranthene 50 μg/mL
    Fluorene 100 μg/mL
    Indeno[1,2,3-cd]pyrene 50 μg/mL
    Naphthalene 500 μg/mL
    Phenanthrene 40 μg/mL
    Pyrene 100 μg/mL
    See All (16)

Signal Word

Danger

Flash Point(F)

closed cup

Flash Point(C)

closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service