Skip to Content
Merck
All Photos(2)

Key Documents

31544

Supelco

Benfuracarb

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30N2O5S
CAS Number:
Molecular Weight:
410.53
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CCOC(=O)CCN(SN(C)C(=O)Oc1cccc2CC(C)(C)Oc12)C(C)C

InChI

1S/C20H30N2O5S/c1-7-25-17(23)11-12-22(14(2)3)28-21(6)19(24)26-16-10-8-9-15-13-20(4,5)27-18(15)16/h8-10,14H,7,11-13H2,1-6H3

InChI key

FYZBOYWSHKHDMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benfuracarb belongs to the benzofuranyl methylcarbamate insecticide group of pesticides used to control insect pests in citrus, maize, sugar beet and vegetables.

Application

Benfuracarb may be used as a reference standard for the determination of the analyte:
  • In bananas by application of a modified QuEChERS procedure followed by ultra high performance liquid chromatography coupled to tandem mass spectrometry (UHPLC-MS/MS).
  • In honeybees, honey and bee pollen by liquid chromatography coupled to tandem mass spectrometry.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pesticide residues in honeybees, honey and bee pollen by LC- MS/MS screening: reported death incidents in honeybees.
Kasiotis KM, et al.
The Science of the Total Environment, 485(2), 633- 642 (2014)
S K Lee et al.
International journal of legal medicine, 112(4), 268-270 (1999-08-05)
We describe here three cases involving acute fatalities due to benfuracarb ingestion and the forensic toxicological implications. Benfuracarb, a carbamate insecticide and its main metabolite carbofuran, were detected using thin layer chromatography (TLC) and gas chromatography/mass spectrophotometry (GC/MS) after extraction
Anjana Srivastava et al.
Pest management science, 67(2), 209-212 (2010-11-13)
Sorption-desorption processes govern the movement of pesticides in soil. These processes determine the potential hazard of the pesticide in a given environment for groundwater contamination and need to be investigated. In the present study, sorption-desorption processes of benfuracarb were investigated
A Maiza et al.
Communications in agricultural and applied biological sciences, 69(3), 257-266 (2005-03-12)
Blatta germanica is the more prevalent cockroach species in Algeria. In the present study, we tested the effect on reproduction in B. germanica of two insect growth regulators, RH-0345, a benzoylhydrazine analogue that mimics the action of 20-hydroxyecdysone, and methoprene
Tatsuhiko Kawamoto et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 19(12), 1605-1610 (2003-12-31)
A simultaneous analytical method was examined for carbofuran and its derivative pesticides in water. Since carbofuran derivatives are hydrolyzed to carbofuran in water, the liquid-liquid extraction method was used to obtain an accurate concentration value. Moreover, since these compounds are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service