Skip to Content
Merck
All Photos(1)

Key Documents

48554

Supelco

N-Nitrosodi-n-propylamine

analytical standard

Synonym(s):

NDPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CCCN(CCC)N=O

InChI

1S/C6H14N2O/c1-3-5-8(7-9)6-4-2/h3-6H2,1-2H3

InChI key

YLKFDHTUAUWZPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Nitrosodi-n-propylamine may be used as an analytical reference standard for the determination of the analyte in wastewater samples gas chromatography—triple quadrupole mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A new method for quantifying N-nitrosamines in wastewater samples by gas chromatography?triple quadrupole mass spectrometry.
Yoon S, et al.
Talanta, 97, 256-261 (2012)
M A Moore et al.
Journal of the National Cancer Institute, 78(2), 289-293 (1987-02-01)
The effects of butylated hydroxyanisole [(BHA) CAS: 25013-16-5], alpha-tocopherol [(TC) CAS: 59-02-9], and carbazole [(CA) CAS: 86-74-8] administered subsequent to a single dose of 2,2'-dioxo-N-nitrosodipropylamine [(DOPN) CAS: 60599-38-4] on the development of putative preneoplastic lesions were investigated in Syrian golden
T Suzuki et al.
Mutation research, 444(2), 259-268 (1999-10-16)
We studied five carcinogens for (a) organ-specific mutagenicity and expression time in the transgenic (TG) mouse mutation assay and (b) clastogenicity in the peripheral blood micronucleus assay in the same mice. Groups of mice were injected intraperitoneally (ip) with N-nitroso-di-n-propylamine
D S Longnecker
Environmental health perspectives, 56, 245-251 (1984-06-01)
Focal proliferative changes in the acinar cells of the pancreas of rats have been induced by several systemically administered carcinogens including azaserine, N-nitrosobis(2-oxopropyl)amine, N-nitroso-(2-hydroxypropyl) (2-oxopropyl)amine, and N delta-(N-methyl-N-nitrosocarbamoyl)-L-ornithine (MNCO). Foci, nodules, and adenomas induced by these carcinogens are usually made
Metabolism of carcinogenic amino derivatives in various species and DNA alkylation by their metabolites.
H A Schut et al.
Drug metabolism reviews, 15(4), 753-839 (1984-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service