Skip to Content
Merck
All Photos(1)

Key Documents

48320-U

Supelco

N-Nitrosodi-n-butylamine solution

certified reference material, 2000 μg/mL in methylene chloride

Synonym(s):

Dibutyl nitrosamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

N(N=O)(CCCC)CCCC

InChI

1S/C8H18N2O/c1-3-5-7-10(9-11)8-6-4-2/h3-8H2,1-2H3

InChI key

YGJHZCLPZAZIHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-Nitrosamines (15 listings): N-Nitrosodi-n-butylamine.
Report on carcinogens : carcinogen profiles, 12, 303-304 (2011-08-24)
N-nitrosodi-n-butylamine.
Report on carcinogens : carcinogen profiles, 11, III196-III197 (2004-01-01)
N-Nitrosodi-n-butylamine.
Report on carcinogens : carcinogen profiles, 10, 175-176 (2004-08-26)
M Hirose et al.
Carcinogenesis, 12(10), 1773-1776 (1991-10-01)
The effects of butylated hydroxyanisole (BHA) pretreatment on subsequent low dose N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) or N,N-dibutylnitrosamine (DBN) treatment on forestomach or esophageal carcinogenesis were investigated in male F344 rats. Groups of animals were pretreated with 2% BHA or basal diet alone
Volatile N-nitrosamines in selected Italian cheeses.
A Dellisanti et al.
Bulletin of environmental contamination and toxicology, 57(1), 16-21 (1996-07-01)

Articles

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service