Skip to Content
Merck
All Photos(1)

Key Documents

73861

Sigma-Aldrich

N-Nitrosodiethylamine

≥99.0% (GC)

Synonym(s):

Diethylnitrosamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NNO
CAS Number:
Molecular Weight:
102.14
Beilstein:
1744991
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥99.0% (GC)

form

liquid

IVD

for in vitro diagnostic use

bp

177 °C (lit.)

density

0.95 g/mL (lit.)

functional group

N-nitroso
amine
nitroso

SMILES string

CCN(CC)N=O

InChI

1S/C4H10N2O/c1-3-6(4-2)5-7/h3-4H2,1-2H3

InChI key

WBNQDOYYEUMPFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

N-Nitrosodiethylamine can be used as a reactant to synthesize:
  • α-Amino oxime derivatives from alkenes via visible-light-induced photoaddition in the presence of HCl[1].
  • Diethylnitrosamine metalloporphyrin complexes from metalloporphyrins in the presence of dichloromethane[2].
N-Nitrosodiethylamine can also be used in the synthesis of 1,1-diethylhydrazine.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Chen et al.
Inorganic chemistry, 37(18), 4677-4688 (2001-10-24)
Diethylnitrosamine reacts with [(TPP)Fe(THF)(2)]ClO(4) (TPP = 5,10,15,20-tetraphenylporphyrinato dianion) in toluene to generate the bis-nitrosamine complex, [(TPP)Fe(Et(2)NNO)(2)]ClO(4), in 96% isolated yield. The related [(TTP)Fe(Et(2)NNO)(2)]SbF(6) (TTP = 5,10,15,20-tetra-p-tolylporphyrinato dianion) complex is prepared in 70% isolated yield via a similar reaction in CH(2)Cl(2).
Changyu Zhu et al.
Journal of hepatology, 74(3), 613-626 (2020-10-11)
The hepatocyte Notch pathway is a pathogenic factor in non-alcoholic steatohepatitis (NASH)-associated fibrosis, but its role in hepatocellular carcinoma (HCC) is less well defined. Herein, we aimed to characterize the molecular and clinical features of Notch-active human HCC, and to
Dilip V Patil et al.
Organic letters, 23(8), 3105-3109 (2021-04-02)
The generation of aminium radical cation species from N-nitrosoamines is disclosed for the first time through visible-light excitation at 453 nm. The developed visible-light-promoted photoaddition reaction of N-nitrosoamines to alkenes was combined with the o-NQ-catalyzed aerobic oxidation protocol of amines
Bo Zhang et al.
Theranostics, 11(10), 4743-4758 (2021-03-24)
Aims: Emerging evidence is demonstrating that rapid regeneration of remnant liver elicited by associating liver partition and portal vein ligation for staged hepatectomy (ALPPS) may be attenuated in fibrotic livers. However, the molecular mechanisms responsible for this process are largely
Koichi Fujisawa et al.
PloS one, 10(6), e0129950-e0129950 (2015-06-25)
Maid is a helix-loop-helix protein that is involved in cell proliferation. In order to further elucidate its physiological functions, we studied Maid activity in two small fish model systems. We found that Maid expression was greatest in zebrafish liver and

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service