Skip to Content
Merck
All Photos(1)

Key Documents

47083-U

Supelco

Formaldehyde solution

stabilized with methanol, ~37 wt. % in H2O, certified reference material

Synonym(s):

Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

CofA

current certificate can be downloaded

autoignition temp.

572 °F

contains

~37% water in methanol as stabilizer

packaging

pkg of 1 mL

concentration

~37 wt. % in H2O

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

1.09 g/mL at 25 °C (lit.)

suitability

suitable for manufacturing use (Laboratory chemicals)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-30°C

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

133.0 °F

Flash Point(C)

56.11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cindy S Ma et al.
The Journal of experimental medicine, 205(7), 1551-1557 (2008-07-02)
Hyper-immunoglobulin E syndrome (HIES) is a primary immune deficiency characterized by abnormal and devastating susceptibility to a narrow spectrum of infections, most commonly Staphylococcus aureus and Candida albicans. Recent investigations have identified mutations in STAT3 in the majority of HIES
Arpeeta Sharma et al.
Diabetes, 64(11), 3937-3950 (2015-06-28)
Patients with diabetes have an increased risk of developing atherosclerosis. Endothelial dysfunction, characterized by the lowered bioavailability of endothelial NO synthase (eNOS)-derived NO, is a critical inducer of atherosclerosis. However, the protective aspect of eNOS in diabetes-associated atherosclerosis remains controversial
G R Moore et al.
Neurology, 55(10), 1506-1510 (2000-11-30)
To determine the pathologic basis of areas not exhibiting signal of the short-T2 component of the T2 relaxation distribution in MS, as studied in formalin-fixed brain. A myelin-specific MRI signal would be of great importance in assessing demyelination in patients
Richard Brad Jones et al.
PLoS pathogens, 10(8), e1004287-e1004287 (2014-08-15)
Resting memory CD4+ T-cells harboring latent HIV proviruses represent a critical barrier to viral eradication. Histone deacetylase inhibitors (HDACis), such as suberanilohydroxamic acid (SAHA), romidepsin, and panobinostat have been shown to induce HIV expression in these resting cells. Recently, it
Stephanie Belloc et al.
Fertility and sterility, 101(6), 1588-1593 (2014-04-03)
To evaluate sperm DNA fragmentation in normozoospermic male partners of couples undergoing infertility evaluation. Retrospective cohort study. Clinical andrology laboratory. A total of 1,974 consecutive normozoospermic men selected from a larger cohort of 4,345 consecutive, nonazoospermic men presenting for infertility

Questions

1–6 of 6 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. Are there any other compounds in Product No. 47083-U, Formaldehyde 37% Wt in Water, except formaldehyde and water?

    1 answer
    1. Cat# 47083-U is a saturated solution of formaldehyde, water, and typically another agent, most commonly methanol. In its typical form, formalin is 37% formaldehyde by weight, 6-13% methanol, and the remainder water.

      Helpful?

  5. Why does Product No. 47083-U, Formaldehyde 37% Wt in Water, contain methanol?

    1 answer
    1. A small percentage of methanol is added to limit the extent of polymerization of the formaldehyde.

      Helpful?

  6. What is the difference between formalin and formaldehyde?

    1 answer
    1. Formaldehyde is a gas that is readily soluble in water. Formalin is the common name for a solution made of 37% formaldehyde, 6-13% methanol, and water. The terms are not interchangeable.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service