Merck
  • Sperm deoxyribonucleic acid damage in normozoospermic men is related to age and sperm progressive motility.
Sperm deoxyribonucleic acid damage in normozoospermic men is related to age and sperm progressive motility.
Fertility and sterility (2014-04-03)
Stephanie Belloc, Moncef Benkhalifa, Martine Cohen-Bacrie, Alain Dalleac, Edouard Amar, Armand Zini
ABSTRACT
To evaluate sperm DNA fragmentation in normozoospermic male partners of couples undergoing infertility evaluation. Retrospective cohort study. Clinical andrology laboratory. A total of 1,974 consecutive normozoospermic men selected from a larger cohort of 4,345 consecutive, nonazoospermic men presenting for infertility evaluation. None. Clinical parameters, conventional semen parameters, and sperm DNA fragmentation assessed by flow cytometry-based TUNEL assay and reported as percent sperm DNA fragmentation (%SDF). The mean (± SD) %SDF and the proportion of men with high %SDF (>30%) were significantly lower in the normozoospermic compared with the entire cohort of 4,345 evaluable infertile men (17.6% ± 10.1% vs. 20.7% ± 12.4% and 11% vs. 20%, respectively). In the group of 1,974 normozoospermic men, %SDF was positively correlated with paternal age (r = 0.17) and inversely correlated with progressive motility (r = -0.26). In the subset of normozoospermic men with sperm parameters above the 50th percentile (≥ 73 × 10(6) sperm/mL, ≥ 55% progressive motility, and ≥ 14% normal forms, World Health Organization 2010 guidelines), 5% (4 of 83) had elevated %SDF (>30%). In this large cohort of normozoospermic men presenting for infertility evaluation, DNA fragmentation level is related to sperm motility and paternal age, and 11% of these men have high levels of sperm DNA fragmentation. Furthermore, the data indicate that a nonnegligible proportion (5%) of normozoospermic men with high-normal sperm parameters may also have significant sperm DNA fragmentation.
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O