Skip to Content
Merck
All Photos(1)

Key Documents

ICS045

Supelco

Formaldehyde Standard

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

manufacturer/tradename

RTC ICS-045-100

concentration

1000 μg/mL in H2O

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application


  • Commercial formaldehyde standard for mass calibration in mass spectrometry.: This study evaluates the use of a commercial formaldehyde standard in mass spectrometry for precise mass calibration, emphasizing its role in enhancing measurement accuracy in pharmaceutical research and biochemistry labs (Lyczko et al., 2015).

  • Honey as a substitute for formalin: This research investigates honey as a potential substitute for formaldehyde in biochemistry applications, comparing its effects on tissue preservation and staining qualities, which may influence biochemistry assays and lab practices (Ozkan et al., 2012).

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service