Skip to Content
Merck
All Photos(1)

Key Documents

46853-U

Supelco

EPA 8270 LCS Mix 1

100 μg/mL each component in acetone: methylene chloride (9:1), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 8270

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 25 mL

concentration

100 μg/mL each component in acetone: methylene chloride (9:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

General description

EPA 8270 LCS Mix 1 is utilized for identifying and quantifying semivolatile organic compounds on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites. The pre-requisite for EPA 8270 method is the use of gas chromatography with a selective detector or gas chromatography/mass spectrometry.
US EPA 8000 Series methods are analytical methods for identifying and quantifying halogenated and aromatic volatile organic compounds (VOCs) on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Aniline
    Anthracene
    Azobenzene
    Benz[a]anthracene
    Benzo[b]fluoranthene
    Benzo[k]fluoranthene
    Benzo[ghi]perylene
    Benzoic acid
    Benzo[a]pyrene
    Benzyl alcohol
    Benzyl butyl phthalate
    Bis(2-chloroethoxy)methane
    Bis(2-chloroethyl) ether
    Bis(2-ethylhexyl) adipate
    Bis(2-ethylhexyl) phthalate
    4-Bromodiphenyl ether
    Carbazole
    4-Chloroaniline
    4-Chlorodiphenyl ether
    Bis-(2-chloroisopropyl) ether
    4-Chloro-3-methylphenol
    2-Chloronaphthalene
    2-Chlorophenol
    Chrysene
    m-Cresol
    p-Cresol
    Dibenz[a,h]anthracene
    Dibenzofuran
    Dibutyl phthalate
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    3,3′-Dichlorobenzidine
    2,4-Dichlorophenol
    Diethyl phthalate
    2,4-Dimethylphenol
    Dimethyl phthalate
    1,2-Dinitrobenzene
    1,3-Dinitrobenzene
    1,4-Dinitrobenzene
    2,4-Dinitrophenol
    2,4-Dinitrotoluene
    2,6-Dinitrotoluene
    Di-n-octyl phthalate
    Fluoranthene
    Fluorene
    Hexachlorobenzene
    Hexachloro-1,3-butadiene
    Hexachlorocyclopentadiene
    Hexachloroethane
    Indeno[1,2,3-cd]pyrene
    Isophorone
    2-Methyl-4,6-dinitrophenol
    1-Methylnaphthalene
    2-Methylnaphthalene
    o-Cresol
    Naphthalene
    2-Nitroaniline
    3-Nitroaniline
    4-Nitroaniline
    Nitrobenzene
    2-Nitrophenol
    4-Nitrophenol
    N-Nitrosodimethylamine
    N-Nitrosodiphenylamine
    N-Nitrosodi-n-propylamine
    Pentachlorophenol
    Phenanthrene
    Phenol
    Pyrene
    Pyridine
    2,3,4,6-Tetrachlorophenol
    2,3,5,6-Tetrachlorophenol
    1,2,4-Trichlorobenzene
    2,4,5-Trichlorophenol
    2,4,6-Trichlorophenol
    See All (78)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-2.2 °F

Flash Point(C)

-19 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A four dimensional separation method based on continuous heart-cutting gas chromatography with ion mobility and high resolution mass spectrometry
Lipok C, et al.
Journal of Chromatography A, 1536, 50-57 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service