Skip to Content
Merck
All Photos(7)

Key Documents

XNAB

Sigma-Aldrich

REDExtract-N-Amp Blood PCR Kit

sufficient for 100 extractions, sufficient for 100 amplifications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

usage

sufficient for 100 amplifications
sufficient for 100 extractions
sufficient for 100 reactions

feature

dNTPs included
hotstart

technique(s)

PCR: suitable

color

red

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The REDExtract-N-Amp Blood PCR Kits contain all of the reagents necessary to rapidly extract host genomic DNA from whole blood and amplify targets of interest by direct PCR (polymerase chain reaction). This novel extraction system eliminates the need for any type of purification, organic extraction, centrifugation, heating, filtration or alcohol precipitation. The kit also includes a PCR ReadyMix, specially formulated for amplification directly from the extract. This formulation uses an antibody based hot start, for specific amplification. The REDExtract-N-Amp Blood PCR Kit contains a tracking dye that allows for convenient direct loading of PCR reactions onto agarose gels for analysis.

Application

REDExtract-N-Amp Blood PCR Kit has been used to extract genomic DNA from blood samples, peripheral blood leukocytes, endocervical swab eluates of mice and parasites. It has also been used to perform polymerase chain reaction (PCR) using oligonucleotide primers.

Features and Benefits

  • Efficient 8 minute prep allows greater speed and throughput
  • No need for organic extraction, centrifugation or alcohol precipitation
  • Hot start antibody for highly specific PCR amplification of genomic DNA
  • Can be used with whole blood or blood cards
  • Extract stable at 4 °C for at least 6 months
  • Compatible with any format (single tube, 96-well, etc.)
  • Simple, 3 step procedure with no special equipment required
  • REDExtract-N-Amp requires no loading buffers or tracking dyes required for gel analysis
  • PCR products are suitable for direct cloning and sequencing

Principle

Genomic DNA is extracted from 10 μl of whole blood by simply adding the Extraction Solution and incubating for 5 minutes at room temperature. The Neutralization Solution is added to the extract to counteract inhibitory substances prior to PCR. A portion of the DNA extract is then added to the specially formulated PCR ReadyMix.

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.
Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • L3289Lysis Solution for BloodSDS

  • N9784Neutralization Solution for BloodSDS

  • P8240REDExtract-N-Amp PCR ReadyMix for BloodSDS

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivier A E Sparagano et al.
Annals of the New York Academy of Sciences, 1081, 505-508 (2006-12-01)
Our study is based on the collection of blood and ticks from sheep in Iran and Italy. Polymerase chain reaction (PCR) testing was performed to target the 18S rRNA gene and RLB was performed using previously published probes. In Italy
Lung-Cheng Lin et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 17(6), 1460-1469 (2008-06-19)
The urinary benzene metabolites, trans, trans-muconic acid (ttMA) and S-phenylmercapturic acid (SPMA), are widely used as benzene exposure biomarkers. The influence of the glutathione S-transferase (GST) genetic polymorphism on the excretion levels of urinary ttMA and/or SPMA has been investigated.
Anna K Nowińska et al.
Molecular vision, 17, 2333-2342 (2011-09-17)
To analyze genotype-phenotype correlation in patients originating from Polish population with the transforming growth factor beta induced (TGFBI) corneal dystrophies. Sixty affected and 31 unaffected individuals from 15 unrelated Polish families were included in the study. The clinical diagnosis was
Kosei Moriyama et al.
Microbiology and immunology, 52(7), 375-382 (2008-08-01)
Bacterial 16S ribosomal RNA genes (rDNA) were detected in blood samples from two healthy individuals by PCR under conditions involving 30 cycles that did not produce any visible products from negative control saline. Even from control samples, PCR involving 35-40
Slawomir J Teper et al.
Molecular vision, 16, 2598-2604 (2010-12-15)
To determine whether gene polymorphisms of the major genetic risk factor for age-related macular susceptibility 2 (ARMS2 A69S) and the complement factor H Y402H influence the response to a variable-dosing treatment regimen with ranibizumab for age-related macular degeneration. This prospective

Articles

Extract-N-Amp simplifies DNA extraction and amplification, enabling PCR directly from FTA blood cards, reducing variability.

Extract-N-Amp simplifies DNA extraction and amplification, enabling PCR directly from FTA blood cards, reducing variability.

Extract-N-Amp simplifies DNA extraction and amplification, enabling PCR directly from FTA blood cards, reducing variability.

Extract-N-Amp simplifies DNA extraction and amplification, enabling PCR directly from FTA blood cards, reducing variability.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service