Skip to Content
Merck
All Photos(4)

Key Documents

X3876

Sigma-Aldrich

Xylanase from Trichoderma viride

greener alternative

lyophilized powder, 100-300 units/mg protein

Synonym(s):

1,4-β-D-Xylanxylanohydrolase, endo-1,4-β-Xylanase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

fungus (Trichoderma viride)

Quality Level

form

lyophilized powder

specific activity

100-300 units/mg protein

mol wt

22 kDa

composition

Protein, ~50% biuret

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

foreign activity

β-glucosidase and β-xylosidase ≤0.1%
cellulase <0.2%

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles and Enzymes for Alternative Energy Research
Xylanase from Trichoderma viride was shown to have molecular weight of 22 kDa with a Pi of 9.3.
Xylanase is a hemicellulolytic enzyme. It is synthesized by microorganisms like bacteria, yeast and fungi.

Application

Xylanase from Trichoderma viride has been used as a component of an enzyme mixture for the hydrolysis of hemicellulose-rich solution (autohydrolysate). It has also been used as a cell-wall degrading enzyme to determine the efficiency of p-coumaryl esterase to release p-coumaroyl and feruloyl group.

Biochem/physiol Actions

Xylanase (endo-1,4-β-Xylanase) is involved in the hydrolysis of xylan. It has a wide range of applications in industrial processes such as, bioleeching of craft pulp in paper industry and production of hydrolysates from agro-industrial wastes. Xylanase is also used in nutritional enhancement of lignocellulosic feed and in clarification of juice and wines.

Unit Definition

One unit will liberate 1 μmole of 4-nitrophenol from 4-nitrophenol-xylan per min at pH 4.5 at 30 °C.

Physical form

Contains sorbitol and sodium acetate buffer salts

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meenakshi Goyal et al.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology], 39(3), 535-541 (2008-07-01)
In the present study, cultural and nutritional conditions for enhanced production of xylanase by a local soil isolate of Trichoderma viride, using various lignocellulosic substrates in submerged culture fermentation have been optimized. Of the lignocellulosics used, maize straw was the
M L T M Polizeli et al.
Applied microbiology and biotechnology, 67(5), 577-591 (2005-06-10)
Xylan is the principal type of hemicellulose. It is a linear polymer of beta-D-xylopyranosyl units linked by (1-4) glycosidic bonds. In nature, the polysaccharide backbone may be added to 4-O-methyl-alpha-D-glucuronopyranosyl units, acetyl groups, alpha-L-arabinofuranosyl, etc., in variable proportions. An enzymatic
Agnieszka Wikiera et al.
Molecules (Basel, Switzerland), 26(5) (2021-04-04)
The biological activity of apple pectin extracted conventionally or enzymatically using endo-xylanase and endo-cellulase, was tested in vitro. The analyses were performerd in tetraplicates and the statistical significance of the differences were assessed using ANOVA, Tukey post hoc and LSD
Low-molecular-weight xylanase from Trichoderma viride
Ujiie, M.
Applied and Environmental Microbiology, 57, 1860-1862 (1994)
Saleh Sabiha-Hanim et al.
Bioresource technology, 102(2), 1234-1239 (2010-08-28)
Oil palm (Elaeis guineensis Jacq.) is one of the most important commercial crops for the production of palm oil, which generates 10.88 tons of oil palm fronds per hectare of plantation as a by-product. In this study, oil palm frond

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service