Skip to Content
Merck
All Photos(4)

Documents

H2125

Sigma-Aldrich

Hemicellulase from Aspergillus niger

greener alternative

powder, 0.3-3.0 unit/mg solid (using a β-galactose dehydrogenase system and locust bean gum as substrate)

Synonym(s):

cellulase, mannanase, xylanase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

specific activity

0.3-3.0 unit/mg solid (using a β-galactose dehydrogenase system and locust bean gum as substrate)

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles and Enzymes for Alternative Energy Research.

Application

Hemicellulase from Aspergillus niger has been used in enzyme digestions.

Biochem/physiol Actions

Hemicellulase is a group of enzymes that catalyzes the breakdown of heteropolysaccharides. It includes glucanases that act on glucan. Similarly, xylanases and mannanases breakdown xylan and mannan respectively. Hemicellulase is prevalent among various bacteria, fungi and plants.

Quality

An undefined mixture of glycolytic enzymes usually containing xylanase, mananase and other activities.

Unit Definition

One unit will produce a relative fluidity change of 1 per 5 minutes using locust bean gum as substrate at pH 4.5 at 40 °C

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Domain-specific mechanosensory transmission of osmotic and enzymatic cell wall disturbances to the actin cytoskeleton
Wojtaszek P, et al.
Protoplasma, 230(3-4), 217-230 (2007)
Handbook of Plant-Based Biofuels, 147-147 (2008)
Alondra M Idrovo Encalada et al.
Food chemistry, 289, 453-460 (2019-04-09)
Carrot residues were upgraded as pectin-enriched fractions (PEFs) useful for functional food formulation due to co-extracted antioxidants (α- and β-carotenes, lutein, α-tocopherol), and gelling effect. High power ultrasound (US)-enzyme assisted extraction was applied for efficiency and sustainability. Carrot powder (CP)
Yanhua Dou et al.
Molecules (Basel, Switzerland), 26(23) (2021-12-11)
Fungal pathogens have evolved combinations of plant cell-wall-degrading enzymes (PCWDEs) to deconstruct host plant cell walls (PCWs). An understanding of this process is hoped to create a basis for improving plant biomass conversion efficiency into sustainable biofuels and bioproducts. Here
Ram Kishor Yadav et al.
Methods in molecular biology (Clifton, N.J.), 959, 235-245 (2013-01-10)
Shoot apical meristems (SAMs) of higher plants harbor a set of stem-cells and provide cells for the development of all the above-ground biomass of plants. Most of the important pattern formation events such as maintenance of stem-cell identity, specification and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service