X2753
Xylanase
powder, ≥2500 units/g, recombinant, expressed in Aspergillus oryzae
Synonym(s):
Pentopan Mono BG®
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Recommended Products
recombinant
expressed in Aspergillus oryzae
form
powder
specific activity
≥2500 units/g
greener alternative product characteristics
Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
storage temp.
2-8°C
General description
Purified endo-β−(1→4)-xylanase from Thermomyces lanuginosus produced by submerged fermentation of a genetically modified Aspergillus oryzae microorganism
Studies show that the signal sequence found in the xylanase gene is critical for protein production in E. coli. Loss of this sequence resulted in a significant reduction in expression.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles and Enzymes for Alternative Energy Research
Xylanase catalyses the degradation of xylan into xylooligosaccharides and D-xylose. It also breaks down hemicellulose. Xylanase is used for the production of biofuels, such as ethanol and xylitol. It improves the quality of bread and juices. Xylanase is used in biobleaching process and decreases environmental pollution.
Application
Xylanase has been used:
- in the isolation of hemicellulose and production of oligosaccharides
- in viscosimetric assay
- in the comparison of commercial enzymes for the aqueous enzymatic extraction of corn oil from corn germ
- in the hydrolytic degradation of xylan
Other Notes
View more information on enzymes for complex carbohydrate analysis at www.sigma-aldrich.com/enzymeexplorer
Legal Information
A product of Novozyme Corp.
Pentopan is a registered trademark of Novozymes Corp.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Microbial xylanases and their industrial application in pulp and paper biobleaching: a review
Walia A, et al.
3 Biotech, 7(1), 11-11 (2017)
Xylan-binding xylanase Xyl30 from Streptomyces avermitilis: cloning, characterization, and overproduction in solid-state fermentation
Hernandez A, et al.
International Microbiology : The Official Journal of the Spanish Society for Microbiology (2008)
Effect of Signal Sequence on the β-xylanase from Thermomyces lanuginosus SKR Expression in Escherichia coli
Khucharoenphaisan , K.
Biotechnology, 10, 209-214 (2011)
Shuangxi Nie et al.
Carbohydrate polymers, 181, 1136-1142 (2017-12-20)
In the present study, cellulose nanofibrils (CNF) were produced from unbleached eucalyptus pulp, and the effect of enzymatic treatment on the properties of CNF was studied. Moreover, the mechanism of enzymatic treatment on the dispersion and film properties of CNF
Svini Dileepa Marasinghe et al.
Microbial cell factories, 20(1), 129-129 (2021-07-10)
Xylanase-containing enzyme cocktails are used on an industrial scale to convert xylan into value-added products, as they hydrolyse the β-1,4-glycosidic linkages between xylopyranosyl residues. In the present study, we focused on xynS1, the glycoside hydrolase (GH) 11 xylanase gene derived
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service