Skip to Content
Merck
All Photos(1)

Key Documents

W2751

Plastic square weighing boats

size 43 mm × 43 mm × 9 mm

Synonym(s):

Plastic weights, Weighing trays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.71
Pricing and availability is not currently available.

material

polystyrene

packaging

case of 4000 ea (Internal packs of 500)

manufacturer/tradename

Sigma-Aldrich

dish L × W

43 mm × 43 mm

size

43 mm × 43 mm × 9 mm

Looking for similar products? Visit Product Comparison Guide

General description

Can be used for liquid or solid samples. The smooth rounded sidewall surface allows easy removal of samples.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service