Key Documents
Z239119
Sigma-Aldrich
White disposable polystyrene pour-boats
W × L × H 40 mm × 31 mm × 8 mm
About This Item
UNSPSC Code:
41100000
NACRES:
NB.71
material
oblong
polystyrene
packaging
pack of 250 ea
manufacturer/tradename
Sigma-Aldrich
W × L × H
40 mm × 31 mm × 8 mm
dish L × W
31 mm × 40 mm
General description
Weighing dish designed to facilitate dispensing and pouring. Smooth uniform walls, flat bottom, usable to 200°F (93°C).