Skip to Content
Merck
All Photos(6)

Documents

U3254

Sigma-Aldrich

Monoclonal Anti-Uvomorulin/E-Cadherin antibody produced in rat

clone DECMA-1, ascites fluid, buffered aqueous solution

Synonym(s):

Anti-E-Cadherin, Anti-LCAM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

DECMA-1, monoclonal

form

buffered aqueous solution

contains

15 mM sodium azide

species reactivity

bovine, human, canine, mouse

technique(s)

immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
indirect immunofluorescence: 1:1,600 using cultured MDCK cells
microarray: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDH1(999)
mouse ... Cdh1(12550)

General description

Monoclonal Anti-Uvomorulin/E-Cadherin (rat IgG1 isotype) is derived from the DECMA-1 hybridoma, produced by the fusion of rat myeloma cells and splenocytes from an immunized Lou rat. Uvomorulin protein was initially identified in embryonal carcinoma and is identical to E-Cadherin, liver-cell adhesion molecules (L-CAM), Cell CAM 80/120, and Activity-regulated cytoskeleton-associated protein 1 (ARC-1), each of which have been characterized in different systems. Uvomorulin/E-Cadherin has been characterized as a 120 kDa cell surface glycoprotein from which an 84 kDa fragment can be released by trypsin digestion in the presence of Ca2+.

Specificity

Monoclonal Anti-Uvomorulin/E-Cadherin was selected against the mouse cell adhesion molecule uvomorulin/E-Cadherin. The antibody localizes the cell surface glycoprotein uvomorulin/E-cadherin that has been found to be identical to L-CAM, Cell CAM 80/120, and ARC-1. It blocks both the aggregation of mouse embryonal carcinoma cells and the compaction of pre-implantation embryos. The antibody disrupts confluent monolayers of Madin-Darby canine kidney (MDCK) epithelial cells. In indirect immunofluorescent staining of MDCK cells grown in culture, the antibody shows strong staining on the membrane of adjacent cells, after treatment with 0.5% Triton-X 100.
The antibody localizes the cell surface glycoprotein uvomorulin/E-cadherin that has been found to be identical to L-CAM, Cell CAM 80/120, and ARC-1. The antibody may be used for studies of embryonal development, cell-cell interactions of cultured cells, and localization of uvomorulin/E-cadherin in immunoblotting or immunohistochemical assays.

Immunogen

mouse embryonal carcinoma cell line PCC4 Aza R1.

Application

Monoclonal Anti-Uvomorulin/E-Cadherin has been used in immunofluorescence, immunoblotting, immunoprecipitation, immunohistochemistry, macromolecule permeability assay and agglomeration of two embryoid body assay.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria P Gavilan et al.
PLoS biology, 13(3), e1002087-e1002087 (2015-03-13)
Epithelial morphogenesis involves a dramatic reorganisation of the microtubule cytoskeleton. How this complex process is controlled at the molecular level is still largely unknown. Here, we report that the centrosomal microtubule (MT)-binding protein CAP350 localises at adherens junctions in epithelial
Sylvain Bessonnard et al.
The Journal of cell biology, 210(7), 1185-1197 (2015-09-30)
The first cell differentiation in mammalian embryos segregates polarized trophectoderm cells from an apolar inner cell mass (ICM). This lineage decision is specified in compacted morulae by cell polarization and adhesion acting on the Yes-associated protein in the Hippo signaling
Claudin-3 overexpression increases the malignant potential of colorectal cancer cells: roles of ERK1/2 and PI3K-Akt as modulators of EGFR signaling
de Souza WF, et al.
PLoS ONE, 8(9), e74994-e74994 (2013)
Controlled, scalable embryonic stem cell differentiation culture
Dang SM, et al.
Stem Cells, 22(3), 275-282 (2004)
TGF-beta induced transdifferentiation of mammary epithelial cells to mesenchymal cells: involvement of type I receptors.
Miettinen PJ, et al.
The Journal of Cell Biology, 127(6), 2021-2036 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service