Skip to Content
Merck
All Photos(3)

Documents

A0349

Sigma-Aldrich

Anti-l-Afadin antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~200 kDa

species reactivity

canine, mouse, rat, human

packaging

antibody small pack of 25 μL

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry (frozen sections): 1:1,000 using mouse liver sections.
indirect immunofluorescence: 1:500 using MDCK cells and cultured human HepG2 cells.
microarray: suitable
western blot: 1:2,000 using extract of rat brain

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLLT4(4301)
mouse ... Mllt4(17356)
rat ... Mllt4(26955)

General description

Afadin is a multidomain protein that binds F-actin and E-catenin proteins at cell-cell adheren junctions. I-afadin is a ubiquitously expressed splice variant of afadin that contains PDZ and F-actin binding domains at the C-terminal end. I-afadin is homologous to the human AF-6 protein .

Specificity

Anti-l-Afadin antibody is specific for I-afadin (approximately 200 kDa). It also binds I-afadin in rat, mouse, dog and human.

Immunogen

Synthetic peptide sequence corresponding to amino acid residues 1814-1829 of rat afadin with N-terminal cysteine conjugated to maleimide-activated KLH.

Application

Anti-l-Afadin antibody produced in rabbit has been used in:
  • western blotting
  • immunofluorescence experiments
  • immunocytochemistry
  • immunohistochemistry

Biochem/physiol Actions

AF-6/Afadin regulates the formation of tight and adherens junctions in the cell and its absence in mice leads to embryonic defects and lethality . Afadin is implicated in primary breast cancer and pancreatic cancer. It downregulates the process of migration and metastasis in cancer cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A genome-wide screen identifies conserved protein hubs required for cadherin-mediated cell-cell adhesion
Toret CP, et al.
The Journal of cell biology, 204(2), 265-279 (2014)
Lauren M Popov et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(46), 14337-14342 (2015-10-23)
Staphylococcus aureus is both a transient skin colonizer and a formidable human pathogen, ranking among the leading causes of skin and soft tissue infections as well as severe pneumonia. The secreted bacterial α-toxin is essential for S. aureus virulence in
Afadin: a key molecule essential for structural organization of cell-cell junctions of polarized epithelia during embryogenesis
Ikeda W, et al.
The Journal of cell biology, 146(5), 1117-1132 (1999)
R Prasad et al.
Cancer research, 53(23), 5624-5628 (1993-12-01)
Reciprocal chromosome translocations involving 11q23 are frequently associated with acute leukemias, with the t(4;11) translocation predominating among acute lymphoblastic leukemias, and the t(9;11), t(11;19) and t(6;11) translocations most common among acute myeloid leukemias. In each of these translocations the ALL-1
Afadin cooperates with Claudin-2 to promote breast cancer metastasis
Tabaries S, et al.
Genes & Development, 33(3-4), 180-193 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service