Skip to Content
Merck
All Photos(3)

Key Documents

A3306

Sigma-Aldrich

α-Amylase, heat-stable

greener alternative

solution, for use in Total Dietary Fiber Assay, TDF-100A

Synonym(s):

α-Amylase from Bacillus licheniformis, Heat-Stable Alpha-Amylase, Heat-stable enzyme, 1,4-α-D-Glucan-glucanohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.32

biological source

bacterial (Bacillus sp.)

form

solution

quality

for use in Total Dietary Fiber Assay, TDF-100A

specific activity

20000—60000 U/mL

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

activity assay: suitable

suitability

suitable for hydrolysis, synthesis of oligosaccharides and polysaccharides, and sugar modification

application(s)

food and beverages

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

α-Amylase (1,4-α-d-glucan glucohydrolase) is an endo-acting glucanase. It belongs to the glycoside hydrolase family 13 (GH13). α-Amylase from Bacillus licheniformis NCIB 6346 can maintain >98% of activity after 60 minutes at pH 6.2 at 85 °C.
α-Amylase, an extracellular enzyme, occurs in many natural sources, including animals, plants, and notably in microorganisms, such as various Bacillus species. α-Amylase is well-known as a heat-stable alkaline enzyme.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for "Energy efficiency" and "waste prevention" when used in starch ethanol research. For more information see the article in biofiles.

Application

α-Amylase, heat-stable has been used:
  • as part of the animal feed in the experimental design to study the effect of dietary fiber on reproductive performance of sows during gestation
  • as a supplement to prepare simulated small intestinal fluid (SSIF) and validate the in vitro digestibility by comparing with endogenous small intestinal fluid (ESIF) on different feeds in ducks
  • to perform an in vitro intestinal digestion using raspberry dietary fiber

Biochem/physiol Actions

α-Amylase breaks down starch into sugars, by hydrolysis of the a-(1→4) glucan linkages in polysaccharides of three or more a-(1→4) linked D-glucose units, without hydrolyzing the α-(1→6) bond. It results in the formation of oligosaccharides including maltose, glucose, and alpha limit dextrin.
α-Amylases are predominantly utilized in processes involving starch saccharification and liquefaction, often requiring high-temperature conditions. These enzymes find wide industrial applications across sectors including food, fermentation, textile, paper, detergent, and pharmaceutical industries. In the processed-food industry, amylases play a vital role in baking, brewing, digestive aid preparation, cake production, fruit juice processing, and starch syrup manufacturing. Apart from their role in generating fermentable compounds, α-amylases contribute to anti-staling effects in breadmaking and enhance the softness retention of baked goods, thereby extending the shelf life of these products.

Other Notes

For R&D use only. Not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bonjin Koo et al.
Journal of animal science, 98(5) (2020-04-21)
This study was conducted to investigate the effects of dietary complexity and l-Thr supplementation on energy and nutrient utilization in nursery pigs. Thirty-two nursery pigs (7.23 ± 0.48 kg) were randomly assigned to a 2 × 2 factorial treatment arrangement
M A Khan et al.
Journal of dairy science, 91(3), 1140-1149 (2008-02-23)
Ruminal parameters, rumen development, nutrient digestibilities, and N utilization were estimated in Holstein calves fed starch from different sources. Ground corn, ground barley, ground wheat, and crimped oats were used to formulate 4 isostarch (25% of starter dry matter) pelleted
Elijah G Kiarie et al.
Journal of animal science, 98(6) (2020-05-22)
Indigestible fiber-protein-phytate complexes reduce the feeding value of soy products. We investigated the effects of multienzyme supplement (MES, Victus) on standardized ileal digestibility (SID) of amino acids (AA) and apparent total tract digestibility (ATTD) of energy and minerals in roasted
Jong Woong Kim et al.
Journal of animal science, 98(1) (2020-01-18)
An experiment was carried out to determine energy values of high-protein sunflower meal (HP-SFM) and to compare the energy values of HP-SFM determined using either a phosphorus (P)-deficient basal diet or a P-adequate basal diet. Twenty-four growing barrows were randomly
J P Marden et al.
Journal of dairy science, 91(9), 3528-3535 (2008-09-04)
The objectives of this study were to evaluate the capacity of 2 dietary feed additives, sodium bicarbonate and live yeast Saccharomyces cerevisiae (strain Sc 47), in optimizing ruminal pH in dairy cows and to determine their modes of action. Three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service