Skip to Content
Merck

C8656

Celatom®, acid-washed

for use in Total Dietary Fiber Assay, TDF-100A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12164500
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Level

quality

for use in Total Dietary Fiber Assay, TDF-100A

technique(s)

photometry: suitable

loss

≤1% loss on ignition

color

white

application(s)

food and beverages
general analytical

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

Celatom®, acid-washed, is used to determine the total dietary fiber content of food using a combination of enzymatic and gravimetric methods.

Application

Celatom®, acid-washed, has been used in a filtration procedure to obtain total dietary fiber (TDF)(2), insoluble dietary fiber((IDF), and soluble dietary fiber (SDF)

Legal Information

Celatom is a registered trademark of EP Minerals, LLC


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yong Zhuo et al.
Journal of animal science and biotechnology, 11, 47-47 (2020-05-20)
This study aimed to investigate the impacts of guar gum and cellulose as the source of dietary fiber during gestation on the reproductive performance of sows. A total of 210 sows (parities 3-6) were randomly allocated into six diets (n = 35)
Guodong Sheng et al.
Environmental science & technology, 45(18), 7718-7726 (2011-08-23)
Sequestration of Ni(II) on diatomite as a function of time, pH, and temperature was investigated by batch, XPS, and EXAFS techniques. The ionic strength-dependent sorption at pH < 7.0 was consistent with outer-sphere surface complexation, while the ionic strength-independent sorption
Myroslav Sprynskyy et al.
Journal of hazardous materials, 181(1-3), 700-707 (2010-06-15)
In this work the natural and the surfactant modified diatomite has been tested for ability to remove uranium ions from aqueous solutions. Such controlling factors of the adsorption process as initial uranium concentration, pH, contact time and ionic strength have



Global Trade Item Number

SKUGTIN
C8656-1KG04061833522684
C8656-50G04061835512331
C8656-50G-KC04061833662496