Skip to Content
Merck
All Photos(4)

Documents

T8691

Sigma-Aldrich

Taurine

suitable for cell culture, BioXtra, meets USP testing specifications

Synonym(s):

2-Aminoethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2SO3H
CAS Number:
Molecular Weight:
125.15
Beilstein:
1751215
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Taurine, suitable for cell culture, meets USP testing specifications

Agency

meets USP testing specifications

product line

BioXtra

form

crystalline powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

mp

>300 °C (lit.)

SMILES string

NCCS(O)(=O)=O

InChI

1S/C2H7NO3S/c3-1-2-7(4,5)6/h1-3H2,(H,4,5,6)

InChI key

XOAAWQZATWQOTB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Taurine has been used:
  • as a component of cardiomyocyte dissociation solution
  • as a supplement for modified synthetic oviduct fluid (m-SOF) medium to culture the presumptive zygotes
  • as a constituent of low Ca2+ solution employed in preparation of collagenase B solution for colony dissociation into single cells
  • to assess its effect on development of bovine embryos fertilized with different bull semen during in vitro fertilization
  • as a component of culture media for derivation of cornea organoids

Biochem/physiol Actions

Non-selective endogenous agonist at glycine receptors. Conditionally essential sulfonated amino acid which modulates apoptosis in some cells; functions in many metabolic activities; a product of methionine and cysteine metabolism.
Taurine is predominantly found in the retina and heart and is also found in the brain, intestine, skeletal muscles and kidneys. It controls the concentration of intracellular calcium, protects against ischemia-reperfusion injury and possesses blood pressure-lowering properties. Taurine also plays a vital role in bile formation and fat digestion. Deficiency of taurine is associated with development of anxiety, hyperactivity, epilepsy and depression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scalable expansion of human pluripotent stem cells in suspension culture
Zweigerdt R, et al.
Nature Protocols, 6(5), 689-689 (2011)
Variations in bovine embryo production between individual donors for OPU-IVF are closely related to glutathione concentrations in oocytes during in vitro maturation
Hidaka T, et al.
Theriogenology, 113(2), 176-182 (2018)
Patient-specific induced pluripotent stem cell models: generation and characterization of cardiac cells
Zanella F and Sheikh F
Methods in Molecular Biology, 147-162 (2014)
The Effect of Various Concentrations of Taurine during In vitro Fertilization on the Development of Bovine Embryos Fertilized with Spermatozoa from Three Different Bulls
Tsuzuki Y, et al.
Asian-Australasian Journal of Animal Sciences (2010)
Cornea organoids from human induced pluripotent stem cells
Foster JW, et al.
Scientific Reports, 7 (2017)

Articles

Ultrapure water from Milli-Q® system shown free of BPA, suitable for embryology research, minimizing endocrine disruptor interference.

Ultrapure water from Milli-Q® system shown free of BPA, suitable for embryology research, minimizing endocrine disruptor interference.

Ultrapure water from Milli-Q® system shown free of BPA, suitable for embryology research, minimizing endocrine disruptor interference.

Ultrapure water from Milli-Q® system shown free of BPA, suitable for embryology research, minimizing endocrine disruptor interference.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service