Skip to Content
Merck
All Photos(1)

Documents

PHR1109

Supelco

Taurine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Aminoethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2SO3H
CAS Number:
Molecular Weight:
125.15
Beilstein:
1751215
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1643361

API family

taurine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NCCS(O)(=O)=O

InChI

1S/C2H7NO3S/c3-1-2-7(4,5)6/h1-3H2,(H,4,5,6)

InChI key

XOAAWQZATWQOTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

Taurine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical dosage form by high performance thin layer chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Non-selective endogenous agonist at glycine receptors. Conditionally essential sulfonated amino acid which modulates apoptosis in some cells; functions in many metabolic activities; a product of methionine and cysteine metabolism.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7126 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simultaneous Determination of N-Acetyl Cysteine and Taurine by HPTLC Method in Active Pharmaceutical Ingredient and Pharmaceutical Dosage Form.
Magesh AR and Dhanaraju MD
American Journal of Analytical Chemistry, 8(11), 742-742 (2017)
Marlene Wewalka et al.
The Journal of clinical endocrinology and metabolism, 99(4), 1442-1451 (2014-01-18)
Bile acids (BAs) are newly recognized signaling molecules in glucose and energy homeostasis. Differences in BA profiles with type 2 diabetes mellitus (T2D) remain incompletely understood. The objective of the study was to assess serum BA composition in impaired glucose-tolerant
Mette Møller Handrup et al.
Pediatric blood & cancer, 60(8), 1292-1298 (2013-02-19)
To determine if the catheter lock taurolidine can reduce the number of catheter-related bloodstream infections (CRBSI) in pediatric cancer patients with tunneled central venous catheters (CVC). During a study period of 34 months, 129 newly placed tunneled CVCs in 112
Felizia K Voss et al.
Science (New York, N.Y.), 344(6184), 634-638 (2014-05-03)
Regulation of cell volume is critical for many cellular and organismal functions, yet the molecular identity of a key player, the volume-regulated anion channel VRAC, has remained unknown. A genome-wide small interfering RNA screen in mammalian cells identified LRRC8A as
Tsutomu Suzuki et al.
Wiley interdisciplinary reviews. RNA, 2(3), 376-386 (2011-10-01)
Mitochondrial DNA mutations that cause mitochondrial dysfunction are responsible for a wide spectrum of human diseases, referred to as mitochondrial diseases. Pathogenic point mutations are found frequently in genes encoding mitochondrial (mt) tRNAs, indicating that impaired functioning of mutant mt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service