Skip to Content
Merck
All Photos(1)

Key Documents

T5393

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−TRITC antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (whole molecule)−Tetramethylrhodamine isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

TRITC conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:64

storage temp.

2-8°C

target post-translational modification

unmodified

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Anti-Mouse IgG (whole molecule)-TRITC antibody is specific for mouse IgG and all subclasses, it cross reacts with mouse IgA and IgM. Antibody purification is by ion exchange chromatography after which it is conjugated to crystalline tetramethylrhodamine isothiocyanate (TRITC).
Whole antiserum is fractionated and then further purified by ion exchange chromatography to provide the IgG fraction of antiserum. This fraction is essentially free of other goat serum proteins. The antibody is conjugated to crystalline tetramethylrhodamine isothiocyanate (TRITC) and then further purified to remove free TRITC. Specificity for mouse IgG is determined by Ouchterlony Double Diffusion (ODD). The antibody preparation is specific for mouse IgG and all subclasses, it cross reacts with mouse IgA and IgM. Identity and purity of the antibody is established by immunoelectrophoresis (IEP), prior to conjugation. Electrophoresis of the antibody preparation followed by diffusion against anti-goat IgG and anti-goat whole serum results in single arcs of precipitation in the γ region.

Specificity

The antibody preparation is specific for mouse IgG and all subclasses, it cross reacts with mouse IgA and IgM.

Immunogen

IgG isolated from pooled normal mouse serum

Application

Anti-Mouse IgG (whole molecule)-TRITC antibody may be used for immunofluorescence at a minimum antibody dilution of 1:64 using mouse spleen cells. Antibody dilution range of 1:50 to 1:250 was used for immunofluorescence in various studies.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Qi et al.
Gene therapy, 21(1), 10-18 (2013-10-11)
The use of small-interfering RNA (siRNA) has great potential for the development of drugs designed to knock down the expression of damage- or disease-causing genes. However, because of the high molecular weight and negative charge of siRNA, it is restricted
M B Graeber et al.
Journal of neuroscience research, 21(1), 18-24 (1988-09-01)
Axotomy of the rat facial nerve leads to mitotic divisions of microglial cells without developing into phagocytes. In order to study the functional characteristics of those activated, i.e., proliferating but nonphagocytic, microglia we investigated the expression of monocyte/macrophage antigens by
Katharina Lampichler et al.
Endocrine-related cancer, 22(5), 793-803 (2015-07-30)
The Hedgehog (Hh) pathway is an important regulator of early tissue patterning and stem cell propagation. It was found to be aberrantly activated in numerous types of human cancer and might be relevant in cancer stem cells. The identification of
Masato Naraoka et al.
World neurosurgery, 129, e63-e72 (2019-05-18)
Early brain injury after subarachnoid hemorrhage (SAH), which is considered a main factor leading to poor outcome, is believed to be caused by the increase of intracranial pressure (ICP) and/or the presence of subarachnoid blood clots (SBC) itself. The purpose
Dolores Baksh et al.
Journal of cellular biochemistry, 101(5), 1109-1124 (2007-06-05)
Wnt signaling is involved in developmental processes and in adult stem cell homeostasis. This study analyzes the role(s) of key Wnt signaling mediators in the maintenance and osteogenesis of mesenchymal stem cells (MSCs). We focus specifically on the involvement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service