Skip to Content
Merck
All Photos(1)

Documents

F4049

Sigma-Aldrich

Monoclonal ANTI-FLAG® M2-FITC antibody produced in mouse

clone M2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal ANTI-FLAG® M2 antibody produced in mouse, Anti-ddddk, Anti-dykddddk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.32

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M2, monoclonal

form

buffered aqueous solution

species reactivity

all

concentration

~1 mg/mL

technique(s)

direct immunofluorescence: 10 μg/mL using mammalian cells fixed with methanol:acetone

isotype

IgG1

immunogen sequence

DYKDDDDK

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Anti Flag M2-FITC antibody is used for the detection of Flag fusion proteins.This monoclonal antibody is produced in mouse and covalently conjugated to flourescein isothiocyanate (FITC). The antibody recognizes the FLAG sequence at the N-terminus, Met N-terminus, and C-terminus.Binding is not Calcium dependent. F4049 is an Affinity purified antibody, increasing sensitivity in most applications.

Application

Antibody is suitable for immunofluorescence, immunocytochemistry, immunohistochemistry, immunoblotting, flow cytometry and electron microscopy. Ideal for use when detection of Flag fusion proteins is required in a murine host, where secondary antibodies could cause cross reactivity.

Learn more product details in ourFLAG® application portal.

Physical form

Solution in 10 mM phosphate buffered saline, plus 1% BSA and 0.1% sodium azide, pH 7.4

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-ichi Asaka et al.
Molecular pharmacology, 71(6), 1487-1493 (2007-03-01)
Multidrug and toxin extrusion 1 (MATE1) has been isolated as an H(+)/organic cation antiporter located at the renal brush-border membranes. Previous studies using rat renal brush-border membrane vesicles indicated that cysteine and histidine residues played critical roles in H(+)/organic cation
Pete Stavropoulos et al.
Journal of immunological methods, 362(1-2), 190-194 (2010-08-31)
The African trypanosome (Trypanosoma brucei) is transmitted by the bite of the tsetse vector to the mammalian bloodstream where it exists as a completely extracellular parasite. As a result of this exposure, the parasite elicits a robust immune response that
Martina Podleschny et al.
PloS one, 10(12), e0145169-e0145169 (2015-12-19)
Neural crest cells are a highly migratory pluripotent cell population that generates a wide array of different cell types and failure in their migration can result in severe birth defects and malformation syndromes. Neural crest migration is controlled by various
Jing Yi et al.
PloS one, 8(10), e78144-e78144 (2013-11-10)
LGR4 is an R-spondin receptor with strong positive effect on Wnt signaling. It plays a critical role in development as its ablation in the mouse led to total embryonic/neonatal lethality with profound defects in multiple organs. Haplotype insufficiency of LGR4
Emmanuelle Despras et al.
Nature communications, 7, 13326-13326 (2016-11-05)
Translesion polymerase eta (polη) was characterized for its ability to replicate ultraviolet-induced DNA lesions that stall replicative polymerases, a process promoted by Rad18-dependent PCNA mono-ubiquitination. Recent findings have shown that polη also acts at intrinsically difficult to replicate sequences. However

Articles

Glycans play a key role in protein structure and disease; representation on cell surfaces is the glycome.

Glycans play a key role in protein structure and disease; representation on cell surfaces is the glycome.

Glycans play a key role in protein structure and disease; representation on cell surfaces is the glycome.

Glycans play a key role in protein structure and disease; representation on cell surfaces is the glycome.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service