Skip to Content
Merck
All Photos(1)

Key Documents

F6257

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)–FITC antibody produced in sheep

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Sheep Anti-Mouse IgG (whole molecule)–Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

sheep

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:32

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections
Anti-Mouse IgG (whole molecule)-FITC antibody is specific for all mouse IgG subclasses. Affinity isolated antigen specific antibody is obtained from sheep anti-mouse IgG antiserum by immunospecific purification. The antibody preparation is then conjugated to Sigma Fluorescein Isothiocyanate (FITC), Isomer I.

Immunogen

Purified mouse IgG

Application

Anti-Mouse IgG (whole molecule)-FITC antibody may be used for immunofluorescent labeling of mouse spleen cells at a working antibody dilution of 1:32. For immunofluorescence of human ovarian adenocarcinoma cells stained with anti-integrin primary antibodies a working dilution of 1:50 was used. Immunolocalization of human spermatids was done using antibody dilution of 1:100. This antibody was also used for immunofluorescent tagging of leaf cells of Arabidopsis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lénaïg Louboutin et al.
Journal of fish diseases, 45(5), 707-716 (2022-02-17)
Gilthead seabream represents a species of importance in Mediterranean aquaculture. The larval stage is particularly sensitive and frequently impacted in suboptimal environmental or sanitary conditions. In the present study, investigations were carried out in a seabream hatchery following an unusual
Fiona Ingrao et al.
Vaccines, 9(7) (2021-08-07)
Recombinant Newcastle disease viruses (rNDV) have been used as bivalent vectors for vaccination against multiple economically important avian pathogens. NDV-vectored vaccines expressing the immunogenic H5 hemagglutinin (rNDV-H5) are considered attractive candidates to protect poultry from both highly pathogenic avian influenza
Sapna V Iyer et al.
PloS one, 8(1), e53532-e53532 (2013-01-24)
Breast cancer is a complex disease which cannot be defined merely by clinical parameters like lymph node involvement and histological grade, or by routinely used biomarkers like estrogen receptor (ER), progesterone receptor (PGR) and epidermal growth factor receptor 2 (HER2)
Hong Chen et al.
Journal of parasitology research, 2013, 591520-591520 (2013-07-19)
The in vitro production of gametocytes and oocysts of the apicomplexan parasite genus Eimeria is still a challenge in coccidiosis research. Until today, an in vitro development of gametocytes or oocysts had only been shown in some Eimeria species. For
Jing Ding et al.
Frontiers in microbiology, 8, 2069-2069 (2017-11-23)
The intestinal phase is critical for trichinellosis caused by Trichinella spiralis (T. spiralis), as it determines both process and consequences of the disease. Several previous studies have reported that T. spiralis induces the initial predominance of a Th1 response during

Articles

In the midst of beeping lab timers, presentations and grant deadlines, it is easy to take for granted the quality of lab reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service