Skip to Content
Merck
All Photos(1)

Key Documents

T4827

Sigma-Aldrich

Trandolapril

≥98% (HPLC), white, powder

Synonym(s):

(2S,3aR,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic acid, Mavik

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34N2O5
CAS Number:
Molecular Weight:
430.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: ≥20 mg/mL

originator

Abbott

SMILES string

CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2C3CCCCC3C[C@H]2C(O)=O

InChI

1S/C24H34N2O5/c1-3-31-24(30)19(14-13-17-9-5-4-6-10-17)25-16(2)22(27)26-20-12-8-7-11-18(20)15-21(26)23(28)29/h4-6,9-10,16,18-21,25H,3,7-8,11-15H2,1-2H3,(H,28,29)/t16-,18+,19-,20-,21-/m0/s1

InChI key

VXFJYXUZANRPDJ-WTNASJBWSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

Application

Trandolapril has been used as an angiotensin-converting enzyme (ACE) inhibitor to study its effects on responsiveness of human retinal endothelial cells (HRECs) to vascular endothelial growth factor (VEGF).

Biochem/physiol Actions

Studies have reported that trandolapril inhibits atherosclerosis1 and decreases the occurrence of atrial fibrillation2.
Trandolapril is an ACE inhibitor.
Trandolapril is an ACE inhibitor. Trandolapril differs from other ACE inhibitors in that it has a longer biological half-life and a high degree of lipophilicity.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Trandolapril is soluble in DMSO at a concentration that is greater than or equal to 20 mg/ml.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Piero Ruggenenti et al.
Journal of the American Society of Nephrology : JASN, 23(10), 1717-1724 (2012-09-01)
Micro- or macroalbuminuria is associated with increased cardiovascular risk factors among patients with type 2 diabetes, but whether albuminuria within the normal range predicts long-term cardiovascular risk is unknown. We evaluated the relationships between albuminuria and cardiovascular events in 1208
Marc S Sabatine et al.
Circulation, 125(2), 233-240 (2011-12-20)
Circulating biomarkers can offer insight into subclinical cardiovascular stress and thus have the potential to aid in risk stratification and tailoring of therapy. We measured plasma levels of 4 cardiovascular biomarkers, midregional pro-atrial natriuretic peptide (MR-proANP), midregional pro-adrenomedullin (MR-proADM), C-terminal
Orly Vardeny et al.
The American journal of cardiology, 107(12), 1705-1709 (2011-04-22)
We used data from patients with stable coronary artery disease (CAD) to assess the risk of new-onset diabetes mellitus (NOD) with β blockers and to determine whether angiotensin-converting enzyme (ACE) inhibition would modify this risk. The Prevention of Events with
Torbjørn Omland et al.
Journal of the American College of Cardiology, 61(12), 1240-1249 (2013-02-19)
The aims of this study were to assess the prognostic value of cardiac troponin I levels, measured with a new high-sensitivity assay, in low-risk patients with stable coronary artery disease (CAD) and to contrast its determinants and prognostic merit with
Lorella Lambertucci et al.
Translational research : the journal of laboratory and clinical medicine, 157(6), 348-356 (2011-05-18)
The purpose of this study was to evaluate the effects of antihypertensive drugs on renal hemodynamics in hypertensive patients during an adrenergic activation by mental stress (MS), which induces renal vasoconstriction in healthy subjects. Renal hemodynamics was assessed twice in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service