Skip to Content
Merck
All Photos(1)

Key Documents

L6292

Sigma-Aldrich

Lisinopril

≥98% (HPLC)

Synonym(s):

(S)-1-[N2-(1-carboxy-3-phenylpropyl)-lysyl-proline dihydrate, MK-521

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31O5N3 · 2H2O
CAS Number:
Molecular Weight:
441.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: ≥10 mg/mL
DMSO: ~6.5 mg/mL (with heating and sonicating)

storage temp.

2-8°C

SMILES string

[H]O[H].[H]O[H].NCCCC[C@H](N[C@@H](CCc1ccccc1)C(O)=O)C(=O)N2CCC[C@H]2C(O)=O

InChI

1S/C21H31N3O5.2H2O/c22-13-5-4-9-16(19(25)24-14-6-10-18(24)21(28)29)23-17(20(26)27)12-11-15-7-2-1-3-8-15;;/h1-3,7-8,16-18,23H,4-6,9-14,22H2,(H,26,27)(H,28,29);2*1H2/t16-,17-,18-;;/m0../s1

InChI key

CZRQXSDBMCMPNJ-ZUIPZQNBSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

Application

Lisinopril has been used to study the antifibrotic effect in duchenne muscular dystrophy mice. It has been used to study the changes in renal-structure and -function in TGR(mRen2)27 (transgenic rat harboring the murine Ren-2 gene) rats upon long term darbepoetin α treatment.

Biochem/physiol Actions

Angiotensin converting enzyme (ACE) inhibitor.

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Roos S Frenay et al.
Journal of the renin-angiotensin-aldosterone system : JRAAS, 13(2), 232-238 (2012-01-28)
Erytropoietin (EPO) has cytoprotective and angiogenic properties and has a beneficial effect in ischaemic conditions. Since the development of renal interstitial abnormalities are often associated with ischaemia, we studied the effects of the long-acting EPO analogue darbepoetin alpha (DA) on
Jason V Baker et al.
PloS one, 7(10), e46894-e46894 (2012-10-20)
Treatments that reduce inflammation and cardiovascular disease (CVD) risk among individuals with HIV infection receiving effective antiretroviral therapy (ART) are needed. We conducted a 2 × 2 factorial feasibility study of lisinopril (L) (10 mg daily) vs L-placebo in combination
Jill A Rafael-Fortney et al.
Circulation, 124(5), 582-588 (2011-07-20)
Nearly universal cardiomyopathy in Duchenne muscular dystrophy (DMD) contributes to heart failure and death. Because DMD patients show myocardial fibrosis well before functional impairment, we postulated that earlier treatment using drugs with antifibrotic effect may be beneficial. Three groups of
Nadir Ulu et al.
The Journal of pharmacology and experimental therapeutics, 345(3), 393-403 (2013-03-27)
Transactivation of epidermal growth factor receptor (EGFR) signaling by G protein-coupled receptors has been implicated in several cardiovascular (CV) conditions, including hypertension, heart failure, and cardiac and vascular hypertrophy. However, the therapeutic potential of EGFR inhibition in these conditions is
Linda F Fried et al.
The New England journal of medicine, 369(20), 1892-1903 (2013-11-12)
Combination therapy with angiotensin-converting-enzyme (ACE) inhibitors and angiotensin-receptor blockers (ARBs) decreases proteinuria; however, its safety and effect on the progression of kidney disease are uncertain. Methods We provided losartan (at a dose of 100 mg per day) to patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service