Skip to Content
Merck
All Photos(2)

Key Documents

T4680

Sigma-Aldrich

Terazosin hydrochloride

≥98% (TLC), powder

Synonym(s):

1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(tetrahydro-2-furanyl)carbonyl]piperazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25N5O4 · HCl
CAS Number:
Molecular Weight:
423.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to off-white

solubility

H2O: soluble 19.60-20.40 mg/mL, clear, colorless to faintly yellow
methanol: 20 mg/mL, clear, colorless
ethanol: 4 mg/mL

originator

Abbott

SMILES string

Cl[H].COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)C4CCCO4

InChI

1S/C19H25N5O4.ClH/c1-26-15-10-12-13(11-16(15)27-2)21-19(22-17(12)20)24-7-5-23(6-8-24)18(25)14-4-3-9-28-14;/h10-11,14H,3-9H2,1-2H3,(H2,20,21,22);1H

InChI key

IWSWDOUXSCRCKW-UHFFFAOYSA-N

Gene Information

General description

Terazosin hydrochloride is used to treat hypertension and benign prostatic hyperplasia (BPH). It is metabolised in the liver. It might be associated with dizziness, headache and asthenia. Terazosin hydrochloride is linked to acute generalized exanthematous pustulosis.

Application

Terazosin hydrochloride has been used for orthogonality and similarity analysis using reversed-phase chromatographic studies5. Furthermore, terazosin has also been used for the evaluation of chemometric techniques to determine orthogonality and similarity in chromatographic methods6.

Biochem/physiol Actions

α1-adrenoceptor antagonist.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Terazosin hydrochloride is soluble in water at 19.60 - 20.40 mg/ml and yields a clear, colorless to faint yellow solution. It is also soluble in ethanol at 4 mg/ml. Furthermore, the product is soluble in methanol at 20 mg/ml, with heat as needed, and yields a clear, colorless solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A G Ramage et al.
European journal of pharmacology, 294(2-3), 645-650 (1995-12-29)
The effects of i.v. infusion of the alpha1-adrenoceptor antagonists doxazosin and terazosin (2 mg kg-1 h-1) on spontaneous hypogastric, renal and inferior cardiac nerve activity, spontaneous bladder contractions, blood pressure, heart rate and femoral arterial flow were investigated separately in
A A Hancock et al.
Journal of receptor and signal transduction research, 15(7-8), 863-885 (1995-09-01)
Terazosin and its enantiomers, antagonists of alpha 1-adrenoceptors, were studied in radioligand binding and functional assays to determine relative potencies at subtypes of alpha 1- and alpha 2-adrenoceptors in vitro. The racemic compound and its enantiomers showed high and apparently
E Van Gyseghema et al.
Journal of chromatography. A, 1074(1-2), 117-131 (2005-06-09)
The starting point of this study was a current set of 32 chromatographic systems used to select initial conditions for method development to determine the impurity profile of a drug. The system exhibiting the best selectivity is then selected for
E Van Gyseghem et al.
Journal of pharmaceutical and biomedical analysis, 41(1), 141-151 (2005-12-15)
Several chemometric techniques were compared for their performance to determine the orthogonality and similarity between chromatographic systems. Pearson's correlation coefficient (r) based color maps earlier were used to indicate selectivity differences between systems. These maps, in which the systems were
R McCarty et al.
Hypertension (Dallas, Tex. : 1979), 27(5), 1115-1120 (1996-05-01)
To examine the contribution of the sympathetic nervous system to the development of hypertension, we injected spontaneously hypertensive rat (SHR) pups and normotensive Wistar-Kyoto rat (WKY) pups twice daily with saline (1.0 mL/kg SC) or terazosin (0.5 mg/kg SC), an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service