Skip to Content
Merck
All Photos(3)

Documents

M5625

Sigma-Aldrich

Menadione

crystalline

Synonym(s):

2-Methyl-1,4-naphthoquinone, Vitamin K3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O2
CAS Number:
Molecular Weight:
172.18
Beilstein:
1908453
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic

Assay

≥98.0% (HPLC)

form

crystalline

technique(s)

HPLC: suitable

color

faint green to green-yellow

mp

105-107 °C (lit.)

solubility

oil: soluble

storage temp.

room temp

SMILES string

CC1=CC(=O)c2ccccc2C1=O

InChI

1S/C11H8O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6H,1H3

InChI key

MJVAVZPDRWSRRC-UHFFFAOYSA-N

Gene Information

human ... NQO1(1728)

Looking for similar products? Visit Product Comparison Guide

Application

Menadione has been used for the generation of reactive oxygen species (ROS), followed by flow cytometry analysis. It has also been used in microbiological evaluation, such as to detect fastidious microorganisms.

Biochem/physiol Actions

Menadione (Vitamin K3) is a synthetic analogue of of 1,4-naphthoquinone with a methyl group in the 2-position. Menadione is used as a phosphatase inhibitor and an inhibitor of mitochondrial DNA polymerase γ (pol γ). Menadione can be used as an oxidative injury (free radical generator) inducing agent.
Menadione is an aromatic polycyclic ketone. It serves as a precursor for vitamin K synthesis. Menadione is converted to menaquinone-4, an active metabolite of vitamin K3. Menadione mediates cell death, by inducing the production of reactive oxygen species through the futile redox cycling.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Menadione is a metabolite of oral vitamin K
Thijssen Henk HW, et al.
The British Journal of Nutrition, 95(2), 260-266 (2006)
Menadione triggers cell death through ROS-dependent mechanisms involving PARP activation without requiring apoptosis
Loor G, et al.
Free Radical Biology & Medicine, 49(12), 1925-1936 (2010)
Investigation of cultivable bacteria isolated from longstanding retreatment-resistant lesions of teeth with apical periodontitis
Signoretti Fernanda GC, et al.
Journal of Endodontics, 39(10), 1240-1244 (2013)
Antioxidants can increase melanoma metastasis in mice
Le Gal K, et al.
Science Translational Medicine, 7(308), 308re8-308re8 (2015)
H Thor et al.
The Journal of biological chemistry, 257(20), 12419-12425 (1982-10-25)
The cytotoxic effects of many quinones are thought to be mediated through their one-electron reduction to semiquinone radicals, which subsequently enter redox cycles with molecular oxygen to produce active oxygen species and oxidative stress. The two-electron reduction of quinones to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service