Skip to Content
Merck
All Photos(1)

Key Documents

SRP5127

Sigma-Aldrich

RhoA, His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ARH12, ARHA, RHO12, RHOH12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~23 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RHOA(387)

General description

Ras homolog family member A (RhoA) is a small GTPase which is part of the Rho gene family. The gene encoding it is localized on human chromosome 3p21.31.

Application

RhoA, His tagged human has been used for in vitro kinase assay.

Biochem/physiol Actions

Ras homolog family member A (RhoA) interacts with protein kinases and serves as a target of activated GTPase. RhoA plays an important role in the initial events of cell protrusion. Rho mediates corneal epithelial migration in response to external stimuli by regulating the organization of the actin cytoskeleton. The inhibition of RhoA signaling is required for both vaccinia morphogenesis and virus-induced cell motility. Vaccinia inhibits RhoA signaling using the viral protein F11 and this can enhance the spread of infection. The protein has a role in basal body docking and planar polarization.

Physical form

Supplied in 50mM MOPS, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphodiesterase 5 Inhibition Limits Doxorubicin-induced Heart Failure by Attenuating Protein Kinase G Ia Oxidation*
Oleksandra P
The Journal of Biological Chemistry (2016)
The PRK2 Kinase Is a Potential Effector Target of both Rho and Rac
GTPases and Regulates Actin Cytoskeletal Organization
SYLVIE VINCENT
Molecular and Cellular Biology (1997)
Jin-Hee Kim et al.
The pharmacogenomics journal, 20(4), 601-612 (2020-02-06)
Previously, we identified Ras homologous A (RHOA) as a major signaling hub in gastric cancer (GC), the third most common cause of cancer death in the world, prompting us to rationally design an efficacious inhibitor of this oncogenic GTPase. Here
Activation of RHOA gene transcription in epithelial tumors may be caused by gene amplification and/or demethylation of its promotor region
Molecular Biology (2006)
Connective Tissue Growth Factor in Regulation of RhoA Mediated Cytoskeletal Tension Associated Osteogenesis of Mouse Adipose-Derived Stromal Cells
Yue Xu
PLoS ONE (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service