Skip to Content
Merck
All Photos(1)

Documents

A9041

Sigma-Aldrich

Arg-Gly-Asp-Ser

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H27N7O8
CAS Number:
Molecular Weight:
433.42
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

powder

composition

Peptide content, ~70%

technique(s)

cell culture | mammalian: suitable

storage temp.

−20°C

SMILES string

N[C@@H](CCCNC(N)=N)C(=O)NCC(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CO)C(O)=O

InChI

1S/C15H27N7O8/c16-7(2-1-3-19-15(17)18)12(27)20-5-10(24)21-8(4-11(25)26)13(28)22-9(6-23)14(29)30/h7-9,23H,1-6,16H2,(H,20,27)(H,21,24)(H,22,28)(H,25,26)(H,29,30)(H4,17,18,19)/t7-,8-,9-/m0/s1

InChI key

NNRFRJQMBSBXGO-CIUDSAMLSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Arg-Gly-Asp-Ser

General description

The tetrapeptide Arg-Gly-Asp-Ser (RGDS) is a key component of the cell attachment domain of fibronectin. The RGDS sequence was found initially to promote the attachment of rat kidney fibroblasts (NRKcells) to fibronectin and synthetic fibronectin peptides coupled to protein-coated plastic. Further investigation indicated that the free RGDS peptide inhibited the attachment of NRK cells to fibronectin coated substrates. The RGDS sequence has been shown to occur in several other proteins, such as the λ receptor on E. coli and the Sindbis coat protein. RGDS is also a target sequence for spirochete adherence of the syphilis bacterium Treponema pallidum.

RGDS has been shown to block fibrinogen-induced aggregation of intact erythrocytes and specific binding of fibrinogen to erythrocyte membranes. The effect of RGDS on transforming growth factor ß1 (TGFß1) mRNA expression and secretion in cultured human mesangial cells has been investigated. RGDS has been utilized in a study of integrin-mediated signal transduction in cultured cells from the sponge Suberites domuncula. RGDS has been demonstrated
to mitigate the binding of Mycobacterium tuberculosis to murine alveolar macrophages

Application

Arg-Gly-Asp-Ser has been used:
  • to study its effects on cell attachment in rats
  • to analyse the interaction of fibrinogen with erythrocytes occurs through integrin related receptor
  • to pretreat the cells, to assess the role of integrin in the cell attachment process
  • to test its competition with platelet-secreted, nanosheet-adsorbed proteins for binding to glycoprotein IIIa (GPIIIa)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

This product is soluble in water (1 mg/ml), yielding a
clear, colorless solution.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A few immobilized thrombins are sufficient for platelet spreading
Okamura Y, et al.
Biophysical Journal, 100(2) (2011)
Lian Leng et al.
Advanced materials (Deerfield Beach, Fla.), 24(27), 3650-3658 (2012-06-21)
The one-step, continuous formation of mosaic hydrogel sheets is presented. A microfluidic device allows controllable incorporation of secondary biopolymers within a flowing biopolymer sheet followed by a cross-linking step that retains the microscale composition. Information is encoded; mosaic stiffness and
Integrin-associated protein (CD47) is a putative mediator for soluble fibrinogen interaction with human red blood cells membrane
De OS, et al.
Biochimica et Biophysica Acta, 1818(3), 481-490 (2012)
Integrin and extracellular matrix interactions regulate engraftment of transplanted hepatocytes in the rat liver
Kumaran V, et al.
Gastroenterology, 129(5), 1643-1653 (2005)
Esther Tejeda-Montes et al.
Acta biomaterialia, 8(3), 998-1009 (2011-09-29)
We report on the combination of a top-down and bottom-up approach to develop thin bioactive membrane scaffolds based on functional elastin-like polymers (ELPs). Our strategy combines ELP cross-linking and assembly, and a variety of standard and novel micro/nanofabrication techniques to

Articles

Tissue engineering has become a key therapeutic tool in the treatment of damaged or diseased organs and tissues, such as blood vessels and urinary bladders.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service