Skip to Content
Merck
All Photos(1)

Key Documents

C0887

Sigma-Aldrich

Chloroperoxidase from Caldariomyces fumago

buffered aqueous suspension, 1,000-2,000 units/mg protein (E1%/280)

Synonym(s):

Chloride Peroxidase, Chloride:hydrogen-peroxide oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

fungus (Caldariomyces fumago)

Quality Level

form

buffered aqueous suspension

specific activity

1,000-2,000 units/mg protein (E1%/280)

mol wt

42 kDa

absorbance ratio

RZ ~1.0

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

A useful alternative to lactoperoxidase for 131I ion labeling studies, for bromination of proteins, and for 36Cl labeling of macromolecules in long-term isolation procedures.

Biochem/physiol Actions

Chloroperoxidase (CPO) is a 42,000 Da extracellular heme glycoenzyme containing ferriprotoporphyrin IX as the prosthetic group. CPO is secreted from fungus and exhibits a broad spectrum of chemical reactivities. It is a peroxide-dependent chlorinating enzyme. It also catalyzes peroxidase-, catalase- and cytochrome P450-type reactions of dehydrogenation, H2O2 decomposition and oxygen insertion, respectively. The enzyme has magnetic and spectroscopic properties similar to that of cyctochrome P-450. CPO from the fungus Caldariomyces fumago has the capacity to chlorinate aromatic hydrocarbons, including polycyclic aromatic hydrocarbons (PAHs).

Unit Definition

One unit will catalyze the conversion of 1.0 μmole of monochlorodimedon to dichlorodimedon per min at pH 2.75 at 25 °C in the presence of potassium chloride and H2O2.

Physical form

Purified suspension in 0.1 M sodium phosphate solution, pH approx. 4.5

inhibitor

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

René Ullrich et al.
Applied and environmental microbiology, 70(8), 4575-4581 (2004-08-06)
Agrocybe aegerita, a bark mulch- and wood-colonizing basidiomycete, was found to produce a peroxidase (AaP) that oxidizes aryl alcohols, such as veratryl and benzyl alcohols, into the corresponding aldehydes and then into benzoic acids. The enzyme also catalyzed the oxidation
R Vázquez-Duhalt et al.
Phytochemistry, 58(6), 929-933 (2001-10-31)
Chloroperoxidase from Caldariomyces fumago was able to chlorinate 17 of 20 aromatic hydrocarbons assayed in the presence of hydrogen peroxide and chloride ions. Reaction rates varied from 0.6 min(-1) for naphthalene to 758 min(-1) for 9-methylanthracene. Mono-, di- and tri-chlorinated
Rabindra Kumar Behera et al.
Journal of inorganic biochemistry, 104(11), 1185-1194 (2010-08-17)
The site specific mutants of the thermophilic P450 (P450 175A1 or CYP175A1) were designed to introduce residues that could act as acid-base catalysts near the active site to enhance the peroxidases activity. The Leu80 in the distal heme pocket of
Kelath Murali Manoj et al.
PloS one, 5(5), e10601-e10601 (2010-05-26)
Many heme enzymes show remarkable versatility and atypical kinetics. The fungal extracellular enzyme chloroperoxidase (CPO) characterizes a variety of one and two electron redox reactions in the presence of hydroperoxides. A structural counterpart, found in mammalian microsomal cytochrome P450 (CYP)
Sudeep Kumar Gade et al.
Biochemical and biophysical research communications, 419(2), 211-214 (2012-02-22)
We report that incorporation of very low concentrations of redox protein cytochrome c and redox active small molecule vitamin C impacted the outcome of one-electron oxidations mediated by structurally distinct plant/fungal heme peroxidases. Evidence suggests that cytochrome c and vitamin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service