Skip to Content
Merck
All Photos(1)

Key Documents

SRP3283

Sigma-Aldrich

FETUIN A human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

Alpha-2-HS-glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

≥100 nM

mol wt

45.0-55.0 kDa

packaging

pkg of 50 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AHSG(197)

General description

The gene AHSG (α-2-HS-glycoprotein, also referred to as fetuin-A) is mapped to human chromosome 3q27. Fetuin A/AHSG is a human plasma glycoprotein belonging to the Cystatin family of proteases inhibitors. Fetuin A is highly expressed in embryonic cells and adult hepatocytes, and is expressed to a lesser extent in monocytes/macrophages. The protein has cystatin-like domains, a calcium phosphate-binding site and a TGF (transforming growth factor)-β cytokine-binding motif. Recombinant human fetuin-A is a heterodimeric protein containing a 282 amino acid A chain and a 27 amino acid B chain. Due to glycosylation, recombinant fetuin-A migrates at an apparent molecular weight of 45–55kDa by SDS-PAGE run under nonreducing conditions.

Application

FETUIN A human has been used to stimulate mRNA expression in human monocytic THP1 cells, human in vitro differentiated adipocytes and C57BL/6 mice.

Biochem/physiol Actions

AHSG (α-2-HS-glycoprotein, also referred to as fetuin-A) is a major serum protein component that exerts various calcium dependent physiological activities, and can mediate growth signaling in certain tumor cells. It also can act as a natural antagonist against specific TGF-β (transforming growth factor β) and BMP (bone morphogenetic protein) signaling proteins. High levels of fetuin-A in the serum is associated with mild osteogenesis imperfecta. On the other hand, reduced levels are seen in Paget′s disease. It is predicted as a potential biomarker for the early diagnosis of hypopharyngeal squamous cell carcinoma.

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.8.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fetuin-A induces cytokine expression and suppresses adiponectin production
Hennige AM, et al.
PLoS ONE, 3(3) (2008)
Fetuin-A induces cytokine expression and suppresses adiponectin production
Hennige AM, et al.
Testing, 3(3) (2008)
Cystatin superfamily.
Ochieng J and Chaudhuri G
Journal of Health Care for the Poor and Underserved, 21, 51-70 (2010)
Human fetuin/alpha2HS-glycoprotein level as a novel indicator of liver cell function and short-term mortality in patients with liver cirrhosis and liver cancer.
Kalabay L
European Journal of Gastroenterology & Hepatology, 14, 389-394 (2002)
alpha 2-HS glycoprotein/fetuin, a transforming growth factor-beta/bone morphogenetic protein antagonist, regulates postnatal bone growth and remodeling.
Szweras M
The Journal of Biological Chemistry, 277, 19991-19997 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service