L7759
Lectin from Arachis hypogaea (peanut)
peroxidase conjugate, lyophilized powder
Synonym(s):
PNA, Peanut agglutinin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.54
Recommended Products
biological source
Arachis hypogaea (peanut)
Quality Level
conjugate
peroxidase conjugate
form
lyophilized powder
peroxidase activity
≥20 units/mg protein
technique(s)
western blot: suitable
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
General Western Blot Protocol:
- Glycoprotein sample size: 500ng
- Lectin Concentration: 0.1ug/ml
- Load samples at 500 ng of glycoprotein per lane
- Run 4-20% Bis-Tris SDS page gel
- Transfer gel to a PVDF membrane
- Block membrane for 1 hr at RT with RIPA buffer (R0278 Sigma)
- Incubate HRP lectin at 0.1ug/ml with RIPA buffer for 2 hours at RT
- Wash membrane 5 x 5 minutes with 25ml RIPA buffer
- Detect using chemiluminescent substrate (CPS1-120)
Biochem/physiol Actions
PNA does not agglutinate normal human erythrocytes, but strongly agglutinates neuraminidase treated erythrocytes. PNA has potent anti-T activity similar to the anti-T antibody in human sera. The lectin can be used to distinguish between human lymphocyte subsets.
Unit Definition
One unit will form 1 mg purpurogallin in 20 sec from pyrogallol at pH 6.0 at 20 °C.
Physical form
Contains sodium citrate
Preparation Note
Prepared from peroxidase (P8375) using a modification of a published method, which favors low molecular weight conjugates. Repurified after conjugation by affinity chromatography.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Matthew R Kudelka et al.
Nature methods, 13(1), 81-86 (2015-12-01)
Protein O-glycosylation has key roles in many biological processes, but the repertoire of O-glycans synthesized by cells is difficult to determine. Here we describe an approach termed Cellular O-Glycome Reporter/Amplification (CORA), a sensitive method used to amplify and profile mucin-type
Ting-Hsuan Chen et al.
Methods in molecular biology (Clifton, N.J.), 2248, 139-153 (2020-11-14)
Virus-like particle (VLP) technology is an alternative platform for developing vaccines to combat seasonal and pandemic influenza. Influenza VLPs are non-infectious nanoparticles that can elicit effective vaccine immunogenicity in hosts. B-cell-activating factor (BAFF, or BLyS) and a proliferation-inducing ligand (APRIL)
Ritva Rice et al.
Scientific reports, 7(1), 12063-12063 (2017-09-25)
Ectothermal reptiles have internal pigmentation, which is not seen in endothermal birds and mammals. Here we show that the development of the dorsal neural tube-derived melanoblasts in turtle Trachemys scripta is regulated by similar mechanisms as in other amniotes, but
Anna Iervolino et al.
PloS one, 10(3), e0119142-e0119142 (2015-03-24)
Dicer is a crucial enzyme for the maturation of miRNAs. Mutations in the Dicer gene are highly associated with Pleuro Pulmonary Blastoma-Family Dysplasia Syndrome (PPB-FDS, OMIM 601200), recently proposed to be renamed Dicer syndrome. Aside from the pulmonary phenotype (blastoma)
Dilan Khalili et al.
Journal of innate immunity, 1-15 (2021-05-18)
Fibrotic lesions accompany several pathological conditions, including tumors. We show that expression of a dominant-active form of the Ras oncogene in Drosophila salivary glands (SGs) leads to redistribution of components of the basement membrane (BM) and fibrotic lesions. Similar to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service