Skip to Content
Merck
All Photos(1)

Key Documents

SRP3138

Sigma-Aldrich

PDGF-BB human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Glioma-derived growth factor (GDGF), Osteosarcoma-derived Growth Factor (ODGF), Platelet-Derived Growth Factor-BB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32190102
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1.0-3.0 ng/mL ED50

mol wt

24.3 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PDGFB(5155)

General description

PDGFs (platelet-derived growth factors) are disulfide-linked dimers consisting of two 12.0-13.5kDa polypeptide chains, designated PDGF-A and PDGF-B chains. Two distinct signaling receptors used by PDGFs have been identified and named PDGFR-α and PDGFR-β. PDGFR-α is high-affinity receptor for each of the three PDGF forms. On the other hand, PDGFR-β interacts with only PDGF-BB and PDGF-AB. The gene PDGF-B is mapped to human chromosome 22q13.1. Recombinant human PDGF-BB is a 24.3kDa disulfide-linked homodimer of two B chains (218 total amino acids).

Application

PDGF-BB human has been used in the cell viability assay. It has been used as a stimulator in PDGFRα (platelet-derived growth factor receptor α)-positive HTPCs (human testicular peritubular cells).

Biochem/physiol Actions

The three naturally occurring PDGFs (platelet-derived growth factors); PDGF-AA, PDGF-BB and PDGF-AB, are potent mitogens for a variety of cell types including smooth muscle cells, connective tissue cells, bone and cartilage cells, and some blood cells. The PDGFs are stored in platelet α-granules and are released upon platelet activation. The PDGFs are involved in a number of biological processes, including hyperplasia, chemotaxis, embryonic neuron development, and respiratory tubule epithelial cell development. PDGF-B along with PDGFR-β participates in vascular maturation. It brings smooth muscle cells and pericytes to growing vessels. Heterozygous mutation in PDGF-B is associated with primary familial brain calcification.

Sequence

SLGSLTIAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIEIVRK KPIFKKATVT LEDHLACKCE TVAAARPVT

Physical form

Lyophilized from 10 mM Acetic Acid.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dovitinib preferentially targets endothelial cells rather than cancer cells for the inhibition of hepatocellular carcinoma growth and metastasis.
Chen ZY, et al.
Journal of Translational Medicine, 10, 245-245 (2012)
Low-level genomic instability is a feature of papillary thyroid carcinoma: an array comparative genomic hybridization study of laser capture microdissected papillary thyroid carcinoma tumors and clonal cell lines.
Finn S, et al.
Archives of Pathology & Laboratory Medicine, 131, 65-73 (2007)
Thomas Daubon et al.
Nature communications, 10(1), 1146-1146 (2019-03-10)
We undertook a systematic study focused on the matricellular protein Thrombospondin-1 (THBS1) to uncover molecular mechanisms underlying the role of THBS1 in glioblastoma (GBM) development. THBS1 was found to be increased with glioma grades. Mechanistically, we show that the TGFβ
Arterial smooth muscle cells express platelet-derived growth factor (PDGF) A chain mRNA, secrete a PDGF-like mitogen, and bind exogenous PDGF in a phenotype- and growth state-dependent manner.
Sjolund M, et al.
The Journal of Cell Biology, 106, 403-413 (1998)
Zhi-Yuan Chen et al.
Journal of translational medicine, 10, 245-245 (2012-12-12)
Dovitinib is a receptor tyrosine kinase (RTK) inhibitor targeting vascular endothelial growth factor receptors, fibroblast growth factor receptors and platelet-derived growth factor receptor β. Dovitinib is currently in clinical trials for the treatment of hepatocellular carcinoma (HCC). In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service