Skip to Content
Merck
All Photos(2)

Documents

F0291

Sigma-Aldrich

hBFGF

FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture

Synonym(s):

Fibroblast Growth Factor-Basic human, FGF2, bFGF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

≤-0.6 ng/mL

mol wt

16.0 kDa

packaging

pkg of 4X25 μg
pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1.00 EU/μg

color

white to faint yellow cast

solubility

water: soluble 0.025 mg, clear, colorless to faintly yellow

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FGF2 (2257)

Looking for similar products? Visit Product Comparison Guide

General description

Fibroblast growth factor-basic (FGF-Basic) is found in basement membranes and sub-endothelial extracellular matrix. FGF-Basic is induced early in the development of the embryo where it is necessary (but not sufficient) for pluripotency and self-renewal of embryonic stem cells (ESC) and lineage defined stem cells; wherein it functions to support proliferation and inhibit differentiation. Self-renewal of hESC depends upon activation of the Activin/Nodal/Smad2,3 branch and suppression of the BMP/GDF/Smad5 branch of the TGFβ-family cell signaling program. FGF-Basic is a competence factor for Nodal support of pluripotency. Activin-A is necessary and sufficient to maintain stemness. It induces the expression of both Nodal and FGF-Basic in human ES cells.

Application

Fibroblast growth factor-basic (FGF-Basic) is required for the maintenance of human embryonic stem cells in culture, and various other stem cell lines, such as mesenchymal stem (stroma) cells (MSC).

Physical form

Lyophilized from 0.2 μm filtered solution in 20 mM Tris and 1 M NaCl, pH 7.0, containing 50 μg bovine serum albumin per 1 μg BFGF.

Analysis Note

The bioactivity of FGF-Basic was measured in a fluorometric assay using the redox sensitive dye, Resazurin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunhui Xu et al.
Stem cells (Dayton, Ohio), 23(3), 315-323 (2005-03-08)
Previous studies have shown that prolonged propagation of undifferentiated human embryonic stem cells (hESCs) requires conditioned medium from mouse embryonic feeders (MEF-CM) as well as matrix components. Because hESCs express growth factor receptors, including those for basic fibroblast growth factor
Lei Xiao et al.
Stem cells (Dayton, Ohio), 24(6), 1476-1486 (2006-02-04)
Human embryonic stem cells (hESCs) self-renew indefinitely while maintaining pluripotency. The molecular mechanism underlying hESCs self-renewal and pluripotency is poorly understood. To identify the signaling pathway molecules that maintain the proliferation of hESCs, we performed a microarray analysis comparing an
C Xu et al.
Nature biotechnology, 19(10), 971-974 (2001-10-03)
Previous studies have shown that maintenance of undifferentiated human embryonic stem (hES) cells requires culture on mouse embryonic fibroblast (MEF) feeders. Here we demonstrate a successful feeder-free hES culture system in which undifferentiated cells can be maintained for at least
R Beklem Bostancioglu et al.
Colloids and surfaces. B, Biointerfaces, 155, 415-428 (2017-05-02)
Accelerated Mesenchymal Stem Cells (MSCs) condensation and robust MSC-matrix and MSC-MSC interactions on nano-surfaces may provide critical factors contributing to such events, likely through the orchestrated signal cascades and cellular events modulated by the extracellular matrix. In this study, human
Brad Davidson et al.
Genes & development, 20(19), 2728-2738 (2006-10-04)
Comprehensive gene networks in Ciona intestinalis embryos provide a foundation for characterizing complex developmental processes, such as the initial phases of chordate heart development. The basic helix-loop-helix regulatory gene Ci-Mesp is required for activation of cardiac transcription factors. Evidence is

Articles

Fibroblast growth factors in cell culture and various growth factors for your research

Frequently asked questions about neural stem cells including NSC derivation, expansion and differentiation.

Frequently asked questions about neural stem cells including NSC derivation, expansion and differentiation.

Frequently asked questions about neural stem cells including NSC derivation, expansion and differentiation.

See All

Protocols

A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.

A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.

A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.

A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service