Skip to Content
Merck
All Photos(1)

Key Documents

P4056

Sigma-Aldrich

Platelet-Derived Growth Factor-BB from rat

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

PDGF-BB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Platelet-Derived Growth Factor-BB from rat, PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

recombinant

expressed in E. coli

Quality Level

form

lyophilized powder

potency

1.62 ng/mL EC50

quality

endotoxin tested

mol wt

~30 kDa

packaging

pkg of 5x50 μg
pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

Platelet-derived growth factor (PDGF) is a 30 kDa cationic polypeptide and consists of two disulphide linked chains A and B. PDGF is expressed as hetero and homodimers : PDGF-AB FDGF-AA, PDGF-BB, PDGF-CC and PDGF-DD.

Application

Platelet-Derived Growth Factor-BB from rat has been used as a growth supplement in mouse embryonic fibroblast culture, motoneuron-muscle co-culture and adult spinal cord neurons culture.

Biochem/physiol Actions

Platelet-derived growth factor (PDGF) elicits multifunctional actions with a variety of cells. It is mitogenic to mesoderm-derived cells, such as dermal and tendon fibroblasts, vascular smooth muscle cells, glial cells, and chondrocytes. PDGF is a potent chemoattractant and activator of neutrophils, monocytes, and fibroblasts. PDGF activates smooth muscle relocalization and multiplication. PDGF mediated increased smooth muscle muscle proliferation might be associated with gene regulation. It increases the synthesis of phospholipids, cholesterol esters, glycogen and prostaglandins, and modulates LDL (low density lipoprotein) receptor binding.

Other Notes

Dimer of the B chain of rat PDGF.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 2.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to stimulate 3H-thymidine incorporation in quiescent NR6R-3T3 fibroblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Platelet-Derived Growth Factor?BB Transactivates the Fibroblast Growth Factor Receptor to Induce Proliferation in Human Smooth Muscle Cells.
Millette E, et al.
Trends in Cardiovascular Medicine, 16.1, 25-28 (2006)
Xin Zhou et al.
Current protocols, 2(5), e416-e416 (2022-05-10)
The serine/threonine protein kinase Akt integrates diverse upstream inputs to regulate cell survival, growth, metabolism, migration, and differentiation. Mounting evidence suggests that Akt activity is differentially regulated depending on its subcellular location, which can include the plasma membrane, endomembrane, and
Role of platelets in the development of atherosclerosis.
Quintana L, et al.
Tissue Engineering: Part A, 15.1, 45-54 (2008)
Yuhan Chen et al.
Journal of inflammation research, 14, 2207-2223 (2021-06-04)
Activation of vascular adventitial fibroblasts (VAFs) upon vascular injury contributes greatly to the medial vascular smooth muscle cells (VSMCs) proliferation, migration and the subsequent neointima formation. A number of factors including fibroblast growth factors (FGFs) have been shown to control
Skeletal muscle tissue engineering: A maturation model promoting long-term survival of myotubes, structural development of the excitation?contraction coupling apparatus and neonatal myosin heavy chain expression.
Das M, et al.
Biomaterials, 30.29, 5392-5402 (2009)

Articles

Platelet-Derived Growth Factor is the principal mitogen found in mammalian serum and is released from platelets during clot formation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service