Skip to Content
Merck
All Photos(1)

Key Documents

SML3754

Sigma-Aldrich

SRT2104

≥98% (HPLC)

Synonym(s):

4-Methyl-N-(2-(3-(morpholinomethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)-2-(pyridin-3-yl)thiazole-5-carboxamide, 4-Methyl-N-(2-{3-[(morpholin-4-yl)methyl]imidazo[2,1-b][1,3]thiazol-6-yl}phenyl)-2-(pyridin-3-yl)-1,3-thiazole-5-carboxamide, GSK 2245840B, GSK-2245840B, GSK2245840B, SRT 2104, SRT-2104

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24N6O2S2
CAS Number:
Molecular Weight:
516.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

SRT2104 is an orally active, potent and highly selective activator of the NAD+-dependent deacetylase sirtuin Sirt1 (EC1.5 = 450 nM) with in vivo efficacy in various animal models, including age-related bone/muscle loss (100 mg/kg/d via chow to mice), HD (0.5% in chow to mice), TNF-induced optic nerve degeneration (2-200 pmol via intravitreal injection to rats).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sirt1 activator SRT2104 protects against oxygen-glucose deprivation/reoxygenation-induced injury via regulating microglia polarization by modulating Sirt1/NF-?B pathway
Brain Research, 1753, 147236-147236 (2021)
Sirtuin 1 activator SRT2104 protects Huntington's disease mice
Annals of Clinical and Translational Neurology, 1(12), 1047-1052 (2014)
SRT2104 extends survival of male mice on a standard diet and preserves bone and muscle mass
Aging Cell, 13(5), 787-796 (2014)
The SIRT1 activator SRT2104 promotes BMP9-induced osteogenic and angiogenic differentiation in mesenchymal stem cells
Mechanisms of Ageing and Development, 207, 111724-111724 (2022)
Axonal protection by a small molecule SIRT1 activator, SRT2104, with alteration of autophagy in TNF-induced optic nerve degeneration
Japanese Journal of Ophthalmology, 64(3), 298-303 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service